Invitrogen™

Power Blotter Select Transfer Stacks, PVDF, regular size

Catalog number: PB5310
Have Questions?
Invitrogen™

Power Blotter Select Transfer Stacks, PVDF, regular size

Catalog number: PB5310
Have Questions?
Catalog Number
PB5310
Unit Size
10 stacks
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
40 stacks
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PB531010 stacks
-
PB534040 stacks
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Invitrogen Power Blotter Select Transfer Stacks are used to transfer proteins using the Power Blotter System. They are disposable, pre-assembled stacks that come with an integrated PVDF transfer membrane for dry blotting of proteins from polyacrylamide gels. Each Power Blotter Select Transfer Stack contains filter paper, membrane, and the appropriate cathode and anode buffers incorporated into a solid gel matrix to allow fast, reliable transfer of proteins.

Features of the Power Blotter Select Transfer Stacks:
Rapid transfer—the ready-to-use transfer stack takes little setup time and protein transfers take place in as little as seven minutes
Efficient and even transfer—stacks are made with high-quality membranes that provide high binding efficiency, uniform transfer, and high detection sensitivity
Increased reproducibility—preassembled stacks provide consistent performance

Learn more about the use of the Power Blotter Select Transfer Stacks in Power Blotter System

PVDF membrane
The PVDF membrane (0.2 μm pore size, low fluorescence) in the stack has higher binding capacity than nitrocellulose. The PVDF membrane is pre-activated and ready for use without any pretreatment with alcohols. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, and radiolabeling. The proteins bind to the membrane through hydrophobic interactions. The protein-binding capacity is 240 μg/cm2.

Regular-size transfer stacks
Regular-size Power Blotter Select Stacks (13 cm x 8.3 cm) can be used with all Invitrogen Novex and NuPAGE midi-gels (or equivalent) or two mini-size gels (all Novex, Bolt, and NuPAGE mini-size gels, or equivalent).

Related products
Power Blotter Select Transfer Stacks, PVDF, mini
Power Blotter Select Transfer Stacks, nitrocellulose, mini
Power Blotter Select Transfer Stacks, nitrocellulose, regular size

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Length (Metric)
13 cm
Width (Metric)
8.3 cm
Format
Transfer Stack
Material
PVDF
Pore Size
0.2 μm
Shipping Condition
Room Temperature
Dimensions (LxW)
Regular, 13 cm x 8.3 cm
Product Line
Power Blotter
Sufficient For
10 Blots

Contents & Storage

10 regular-size transfer stacks (13 cm x 8.3 cm)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text