Thermo Scientific™

Pierce™ Premium Grade Sulfo-NHS-LC-Biotin

Catalog number: PG82075
Have Questions?
Thermo Scientific™

Pierce™ Premium Grade Sulfo-NHS-LC-Biotin

Catalog number: PG82075
Have Questions?
Catalog Number
PG82075
Unit Size
100 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PG82075100 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Premium Grade Sulfo-NHS-LC-Biotin is our highest quality formulation of this popular amine-reactive biotinylation reagent, specially characterized for applications where product integrity and risk minimization are paramount.

Features of Premium Grade Sulfo-NHS-LC-Biotin:

Sulfo-NHS-LC-Biotin—popular amine-reactive biotinylation reagent for antibody labeling
High quality—identity and purity confirmed by several tests, including quantitative NMR
Product integrity—enhanced level of testing and characterization compared to standard grade
Lot retention—ample supply of past lots retained to ensure future process testing
Change management—Change Control Notification (CCN) service
Consistent manufacture—batch-specific manufacturing documentation review

Compared to the standard grade product, Premium Grade Sulfo-NHS-LC-Biotin provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines. Specific labeling of cell surface proteins is another common application for these uniquely water-soluble and membrane impermeable reagents.

Related Products
EZ-Link™ Sulfo-NHS-LC-Biotin
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Sulfo-NHS-LC-Biotin
Quantity
100 mg
Product Line
Pierce™
Cell Permeability
Cell-Impermeant
Chemical Reactivity
Amine
Reactive Moiety
Active Ester, Succinimidyl Ester, Sulfo-NHS Ester
Solubility
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye
Biotin
Spacer
Mid-length
Label Type
Biotin & Analogs

Contents & Storage

Upon receipt store desiccated at -20° C. Product is shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text