Thermo Scientific™

Pierce™ Premium Grade Sulfo NHS-SS-Biotin

Catalog number: PG82077
Have Questions?
Thermo Scientific™

Pierce™ Premium Grade Sulfo NHS-SS-Biotin

Catalog number: PG82077
Have Questions?
Catalog Number
PG82077
Unit Size
100 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PG82077100 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific™ Pierce™ Premium Grade Sulfo-NHS-SS-Biotin is our highest quality formulation of this amine-reactive, cleavable biotinylation reagent, specially characterized for applications where product integrity and risk minimization are paramount. Sulfo-NHS-SS-biotin is used for cleavable attachment of biotin to protein and cell surface amines.

• High quality—identity and purity confirmed by multiple tests, including quantitative NMR
• Product integrity—enhanced level of testing and characterization compared to standard grade
• Lot retention—ample supply of past lots retained to help ensure future process testing
• Change management—Change Control Notification (CCN) service
• Consistent manufacture—batch-specific manufacturing documentation review

Compared to the standard grade product, Premium Grade Sulfo-NHS-SS-Biotin provides more clearly defined quality and product support by including increased analytical testing and product characterization, greater batch-specific information and quality assurance review, extensive lot sample retention, and change control notification.

Sulfo-NHS-SS-biotin is a water-soluble, NHS-ester biotinylation reagent with a spacer arm that includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines. This reagent is water-soluble, enabling biotinylation in the absence of organic solvents such as DMSO or DMF. The compound is particularly useful for labeling and purifying cell surface proteins because its sulfonate group prevents it from permeating cell membranes and its cleavable spacer arm enables initially biotinylated proteins to be released from streptavidin affinity columns.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Sulfo NHS-SS-Biotin
Quantity
100 mg
Product Line
Pierce™
Cell Permeability
Cell-Impermeant
Chemical Reactivity
Amine
Reactive Moiety
Active Ester, Succinimidyl Ester, Sulfo-NHS Ester
Solubility
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye
Biotin
Spacer
Mid-length, Cleavable
Label Type
Biotin & Analogs

Contents & Storage

Premium Grade Sulfo-NHS ester of biotin with an ethyl-1,3-dithioproprionate spacer.
Store with desiccant at -20°C.
Sufficient to label a total of 1 g of IgG using typical conditions.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text