Thermo Scientific™
TRACE™ 1310 GC Analyzers for Permanent Gases & Trace Impurities
Catalog number: PGACOCO2001
Have Questions?
Thermo Scientific™
TRACE™ 1310 GC Analyzers for Permanent Gases & Trace Impurities
Catalog number: PGACOCO2001
Have Questions?
Refuse to compromise by choosing from our variety of Thermo Scientific™ TRACE™ 1310 gas chromatography systems for the analysis of industrial gas streams. These systems can be used in laboratories and in pilot/scale-up to at-line environments. Whether you need to analyze permanent gases, hydrogen, low levels of CO/CO2, impurities in chlorine, trace-level impurities in pure gases or a combination of those analyses, there is a system to meet your requirements. TRACE 1310 analyzers for industrial gas streams come pre-configured, factory tested and ready to connect to your lab services.
Catalog Number
PGACOCO2001
Unit Size
Each
Description
Dual-channel gas chromatograph with configured valve oven
Includes
TRACE 1310 GC single TCD detector, single FID detector, auxiliary valve oven with (1) 10-port valves, (1) 6-port valve, 2 columns, and complete documentation package. System is fully assembled and factory tested.
Price (USD)
Full specifications
TypePermanent Gas Analyzer
Detection ModeThermal Conductivity and Flame Ionization
Column Capacity2
DescriptionDual-channel gas chromatograph with configured valve oven
IncludesTRACE 1310 GC single TCD detector, single FID detector, auxiliary valve oven with (1) 10-port valves, (1) 6-port valve, 2 columns, and complete documentation package. System is fully assembled and factory tested.
FunctionAutomated analysis of a gas sample for permanent gases, low levels of CO, CO2, and light hydrocarbons C1-C4
Detector TypeTCD and FID
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Single-channel gas chromatograph with configured valve oven
Includes
TRACE 1310 GC, two TCD detectors, auxiliary valve oven with (2) 10-port valves, (1) 6-port valve, 4 columns, and complete documentation package. System is fully assembled and factory tested.
Price (USD)
Full specifications
TypeSingle Channel Gas Chromatograph with Configured Valve Oven
Detection ModeThermal Conductivity
Column Capacity4
DescriptionSingle-channel gas chromatograph with configured valve oven
IncludesTRACE 1310 GC, two TCD detectors, auxiliary valve oven with (2) 10-port valves, (1) 6-port valve, 4 columns, and complete documentation package. System is fully assembled and factory tested.
FunctionAutomated analysis of impurities in chlorine gas
Detector TypeTCD
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Single-channel gas chromatograph with configured valve oven
Includes
TRACE 1310 GC single FID detector, auxiliary valve oven with (1) 10-port valves, methanizer, 2 columns, and complete documentation package. System is fully assembled and factory tested.
Price (USD)
Full specifications
TypeSingle Channel Gas Chromatograph with Configured Valve Oven
Detection ModeFlame Ionization
Column Capacity2
DescriptionSingle-channel gas chromatograph with configured valve oven
IncludesTRACE 1310 GC single FID detector, auxiliary valve oven with (1) 10-port valves, methanizer, 2 columns, and complete documentation package. System is fully assembled and factory tested.
FunctionAutomated analysis of a gas sample for low level CO2 and CO via methanizer FID
Detector TypeFID
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Dual-channel gas chromatograph with configured valve oven
Includes
TRACE 1310 GC two TCD detector, auxiliary valve oven with (2) 10-port valves, (1) 6-port valve, 4 columns, and complete documentation package. System is fully assembled and factory tested.
Price (USD)
Full specifications
TypePermanent Gas Analyzer
Detection ModeThermal Conductivity
Column Capacity4
DescriptionDual-channel gas chromatograph with configured valve oven
IncludesTRACE 1310 GC two TCD detector, auxiliary valve oven with (2) 10-port valves, (1) 6-port valve, 4 columns, and complete documentation package. System is fully assembled and factory tested.
FunctionAutomated analysis of a gas sample for permanent gases, CO, CO2, and light hydrocarbons C1-C4 and full range analysis of hydrogen content
Detector TypeTCD
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Single-channel gas chromatograph with configured valve oven
Includes
TRACE 1310 GC single TCD detector, auxiliary valve oven with (1) 10-port valves, (1) 6-port valve, 2 columns, and complete documentation package. System is fully assembled and factory tested.
Price (USD)
Full specifications
TypePermanent Gas Analyzer
Detection ModeThermal Conductivity
Column Capacity2
DescriptionSingle-channel gas chromatograph with configured valve oven
IncludesTRACE 1310 GC single TCD detector, auxiliary valve oven with (1) 10-port valves, (1) 6-port valve, 2 columns, and complete documentation package. System is fully assembled and factory tested.
FunctionAutomated analysis of a gas sample for permanent gases, CO, CO2, and light hydrocarbons C1-C4
Detector TypeTCD
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Single-channel gas chromatograph with configured valve oven
Includes
TRACE 1310 GC single PDD detector, auxiliary valve oven with (1) 10-port purged valve, (1) 6-port purged valve, 3 columns, and complete documentation package. System is fully assembled and factory tested.
Price (USD)
Full specifications
TypeSingle Channel Gas Chromatograph with Configured Valve Oven
Detection ModePulsed Discharge
Column Capacity3
DescriptionSingle-channel gas chromatograph with configured valve oven
IncludesTRACE 1310 GC single PDD detector, auxiliary valve oven with (1) 10-port purged valve, (1) 6-port purged valve, 3 columns, and complete documentation package. System is fully assembled and factory tested.
FunctionAutomated analysis of pure He for impurities permanent gases, CO, CO2, and light hydrocarbons C1-C4
Detector TypePDD
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Single-channel gas chromatograph with configured valve oven
Includes
TRACE 1310 GC single PDD detector, oxygen stripper, auxiliary valve oven with (1) 10-port purged valve, (1) 6-port purged valve, (1) 4-port purged valve, 2 columns, and complete documentation package. System is fully assembled and factory tested.
Price (USD)
Full specifications
TypeSingle Channel Gas Chromatograph with Configured Valve Oven
Detection ModePulsed Discharge
Column Capacity2
DescriptionSingle-channel gas chromatograph with configured valve oven
IncludesTRACE 1310 GC single PDD detector, oxygen stripper, auxiliary valve oven with (1) 10-port purged valve, (1) 6-port purged valve, (1) 4-port purged valve, 2 columns, and complete documentation package. System is fully assembled and factory tested.
FunctionAutomated analysis of pure oxygen for impurities permanent gases, CO, CO2, and light hydrocarbons C1-C4
Detector TypePDD
Unit SizeEach
Showing 7 of 7
Catalog NumberSpecificationsUnit SizeDescriptionIncludesPrice (USD)
PGACOCO2001Full specifications
EachDual-channel gas chromatograph with configured valve ovenTRACE 1310 GC single TCD detector, single FID detector, auxiliary valve oven with (1) 10-port valves, (1) 6-port valve, 2 columns, and complete documentation package. System is fully assembled and factory tested.Request A Quote
TypePermanent Gas Analyzer
Detection ModeThermal Conductivity and Flame Ionization
Column Capacity2
DescriptionDual-channel gas chromatograph with configured valve oven
IncludesTRACE 1310 GC single TCD detector, single FID detector, auxiliary valve oven with (1) 10-port valves, (1) 6-port valve, 2 columns, and complete documentation package. System is fully assembled and factory tested.
FunctionAutomated analysis of a gas sample for permanent gases, low levels of CO, CO2, and light hydrocarbons C1-C4
Detector TypeTCD and FID
Unit SizeEach
CL2GAS000001Full specifications
EachSingle-channel gas chromatograph with configured valve ovenTRACE 1310 GC, two TCD detectors, auxiliary valve oven with (2) 10-port valves, (1) 6-port valve, 4 columns, and complete documentation package. System is fully assembled and factory tested.Request A Quote
TypeSingle Channel Gas Chromatograph with Configured Valve Oven
Detection ModeThermal Conductivity
Column Capacity4
DescriptionSingle-channel gas chromatograph with configured valve oven
IncludesTRACE 1310 GC, two TCD detectors, auxiliary valve oven with (2) 10-port valves, (1) 6-port valve, 4 columns, and complete documentation package. System is fully assembled and factory tested.
FunctionAutomated analysis of impurities in chlorine gas
Detector TypeTCD
Unit SizeEach
COCO20010011Full specifications
EachSingle-channel gas chromatograph with configured valve ovenTRACE 1310 GC single FID detector, auxiliary valve oven with (1) 10-port valves, methanizer, 2 columns, and complete documentation package. System is fully assembled and factory tested.Request A Quote
TypeSingle Channel Gas Chromatograph with Configured Valve Oven
Detection ModeFlame Ionization
Column Capacity2
DescriptionSingle-channel gas chromatograph with configured valve oven
IncludesTRACE 1310 GC single FID detector, auxiliary valve oven with (1) 10-port valves, methanizer, 2 columns, and complete documentation package. System is fully assembled and factory tested.
FunctionAutomated analysis of a gas sample for low level CO2 and CO via methanizer FID
Detector TypeFID
Unit SizeEach
H2PGA0202011Full specifications
EachDual-channel gas chromatograph with configured valve ovenTRACE 1310 GC two TCD detector, auxiliary valve oven with (2) 10-port valves, (1) 6-port valve, 4 columns, and complete documentation package. System is fully assembled and factory tested.Request A Quote
TypePermanent Gas Analyzer
Detection ModeThermal Conductivity
Column Capacity4
DescriptionDual-channel gas chromatograph with configured valve oven
IncludesTRACE 1310 GC two TCD detector, auxiliary valve oven with (2) 10-port valves, (1) 6-port valve, 4 columns, and complete documentation package. System is fully assembled and factory tested.
FunctionAutomated analysis of a gas sample for permanent gases, CO, CO2, and light hydrocarbons C1-C4 and full range analysis of hydrogen content
Detector TypeTCD
Unit SizeEach
PGA000010011Full specifications
EachSingle-channel gas chromatograph with configured valve ovenTRACE 1310 GC single TCD detector, auxiliary valve oven with (1) 10-port valves, (1) 6-port valve, 2 columns, and complete documentation package. System is fully assembled and factory tested.Request A Quote
TypePermanent Gas Analyzer
Detection ModeThermal Conductivity
Column Capacity2
DescriptionSingle-channel gas chromatograph with configured valve oven
IncludesTRACE 1310 GC single TCD detector, auxiliary valve oven with (1) 10-port valves, (1) 6-port valve, 2 columns, and complete documentation package. System is fully assembled and factory tested.
FunctionAutomated analysis of a gas sample for permanent gases, CO, CO2, and light hydrocarbons C1-C4
Detector TypeTCD
Unit SizeEach
TIGAHE010013Full specifications
EachSingle-channel gas chromatograph with configured valve ovenTRACE 1310 GC single PDD detector, auxiliary valve oven with (1) 10-port purged valve, (1) 6-port purged valve, 3 columns, and complete documentation package. System is fully assembled and factory tested.Request A Quote
TypeSingle Channel Gas Chromatograph with Configured Valve Oven
Detection ModePulsed Discharge
Column Capacity3
DescriptionSingle-channel gas chromatograph with configured valve oven
IncludesTRACE 1310 GC single PDD detector, auxiliary valve oven with (1) 10-port purged valve, (1) 6-port purged valve, 3 columns, and complete documentation package. System is fully assembled and factory tested.
FunctionAutomated analysis of pure He for impurities permanent gases, CO, CO2, and light hydrocarbons C1-C4
Detector TypePDD
Unit SizeEach
TIGAOX010011Full specifications
EachSingle-channel gas chromatograph with configured valve ovenTRACE 1310 GC single PDD detector, oxygen stripper, auxiliary valve oven with (1) 10-port purged valve, (1) 6-port purged valve, (1) 4-port purged valve, 2 columns, and complete documentation package. System is fully assembled and factory tested.Request A Quote
TypeSingle Channel Gas Chromatograph with Configured Valve Oven
Detection ModePulsed Discharge
Column Capacity2
DescriptionSingle-channel gas chromatograph with configured valve oven
IncludesTRACE 1310 GC single PDD detector, oxygen stripper, auxiliary valve oven with (1) 10-port purged valve, (1) 6-port purged valve, (1) 4-port purged valve, 2 columns, and complete documentation package. System is fully assembled and factory tested.
FunctionAutomated analysis of pure oxygen for impurities permanent gases, CO, CO2, and light hydrocarbons C1-C4
Detector TypePDD
Unit SizeEach
Showing 7 of 7
Ready to Install and Run Samples
  • Fully configured and factory tested before delivery
  • Full documentation set
    • start-up and troubleshooting guide
    • detailed flow and plumbing schematics
    • spare parts list
    • method and factory test results
Built to maximize productivity and uptime
  • Two-oven design: Temperature programmable GC Oven and independent isothermal valve oven can hold up to six packed columns for optimal column management
  • Features Instant Connect detector modules, increasing uptime with detector exchange in minutes
  • Optional third isothermal oven for molecular sieve columns
  • Spacious independent valve oven with easy access to valves, rotors, and actuators
  • Heated sample bulkheads ensure complete sample transfer
  • Optimized system footprint to reduce bench space requirement
  • Permanent Gas - CO/CO2
    • Single channel system for N2, O2, CO, and CO2
    • Single channel for low level CO and CO2
    • Dual channel system for N2, O2, and low-level CO/CO2
    • Dual Channel system for N2, O2, CO, CO2 and H2
  • Trace-level impurities at ppb-ppt in pure gases
    • Impurities in Oxygen - H2, Ar, N2, CO, CO2 and C1-C4
    • Impurities in Helium - H2, Ar, N2, O2, CO, CO2 and C1-C4
    • Impurities in inert, pure gases - H2, Ar, N2, O2, CO, CO2 and C1-C4
  • Impurities in Chlorine Gas
    • Channel 1 - H2
    • Channel 2 - Ar, N2, O2, CO, CO2 and C1-C4