A proven large chamber SEM for rapid materials imaging and analysis
The Thermo Scientific Phenom XL G2 Desktop SEM sets the stage for faster and more efficient quality control and failure analysis.
Its larger stage accommodates samples up to 100 mm x 100 mm, and its automation for routine analytical tasks delivers accurate, reproduceable results with an average time to image of just 60 seconds.
The spacious chamber allows for advanced features like motorized eucentric sample tilting, in situ tensile testing, and electrical feedthrough options. Available argon-compatible models offer an inert environment to protect delicate samples.
The long-life CeB₆ (cerium hexaboride) source gives you outstanding brightness, making it far easier to obtain high-quality images featuring many small details.
The Phenom XL G2 Desktop SEM can also help you streamline your operations by analyzing a greater quantity of samples with less margin for human error. It’s an ideal choice for steel, and equally effective for both routine failure analysis and bespoke research, which is why busy industrial lab and quality managers worldwide continue to rely on this proven all-rounder.  
Phenom XL G2 Desktop SEM overview
Analyze more samples, more efficiently
Expand your research horizons thanks to a larger stage that can accommodate samples up to 100 mm x 100 mm. 
Doing the heavy lifting for you
Automate various stages of your routine workflow, delivering accurate results with an average time to image of 60 seconds.
Making routine analysis more reliable
The SEM’s non-reactive environment enables you to study a wide range of reactive materials. 
Built for your biggest needs
Providing you with the insights you need to discover failures earlier and adjust your process faster.
Phenom XL G2 Desktop SEM resources
Would you like to know more about the Phenom XL G2 Desktop SEM? Dive into a wealth of resources, from eBooks to datasheets. 
Phenom XL G2 Desktop SEM image gallery
See for yourself what this versatile desktop SEM can deliver for you and your team.
For Research Use Only. Not for use in diagnostic procedures.
1x1 image pixel for data collection