PD98059
Product Image
Invitrogen™

PD98059

PD98059 is a potent and selective cell permeable inhibitor of MAP kinase kinase (MEK). It selectively blocks the activation ofRead more
Have Questions?
Catalog NumberQuantity
PHZ11645 mg
Catalog number PHZ1164
Price (USD)
225.00
Each
In stock
Add to cart
Quantity:
5 mg
Price (USD)
225.00
Each
Add to cart
PD98059 is a potent and selective cell permeable inhibitor of MAP kinase kinase (MEK). It selectively blocks the activation of MEK, thereby inhibiting the phosphorylation and the activation of MAP kinase.

Format/Formulation: Lyophilized
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gene ID (Entrez)5604, 5605, 5606, 5607, 5608, 5609, 6416
Inhibitor PropertiesChemical
InhibitorsKinase Inhibitor, Kinase Inhibitor (MAP Kinase Inhibitor)
Molecular Weight (g/mol)267.3 Da
Purification MethodHPLC
Quantity5 mg
Research CategorySignal Transduction
Shipping ConditionDry Ice
Specific Inhibit or TargetMEK (MAP kinase kinase)
SterilityNon-sterile
FormSolid
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link