Search Thermo Fisher Scientific
Catalog Number | Description |
---|---|
PID-10131047C also known as PID-101310 47C | VersaWeigh - Power-User Option Pack |
PID-10131047E also known as PID-101310 47E | VersaWeigh - Reject Integrity Option Pack |
PID-10131047F also known as PID-101310 47F | VersaWeigh - Combination metal detector/checkweigher |
PID-10131047D also known as PID-101310 47D | VersaWeigh - Pharma Option Pack |
PID-10131047 also known as PID-101310 47 | VersaWeigh Checkweigher |
PID-10131047B also known as PID-101310 47B | VersaWeigh - Electrical Option Pack |
PID-10131047A also known as PID-101310 47A | VersaWeigh - IP-66 Option Pack |
Maximize profits and minimize product giveaway with the Thermo Scientific™ Global VersaWeigh™ Checkweigher. This versatile, scalable system is still easy-to-use and maintain but now features more than 10 engineering and functionality improvements including frames designed to reduce food buildup, rapid-release, adjustable conveyors and belts, and high-torque sealed brushless motors for trouble-free operation. The fixed-height Global VersaWeigh is ideal for products up to 10 kilograms (22 pounds) and performs with an accuracy equal to or better than market requirement standards.
The Global VersaWeigh Checkweigher Features: