Search Thermo Fisher Scientific
Catalog Number | Description |
---|---|
PID-10131047E also known as PID-101310 47E | VersaWeigh - Reject Integrity Option Pack |
PID-10131047F also known as PID-101310 47F | VersaWeigh - Combination metal detector/checkweigher |
PID-10131047D also known as PID-101310 47D | VersaWeigh - Pharma Option Pack |
PID-10131047 also known as PID-101310 47 | VersaWeigh Checkweigher |
PID-10131047C also known as PID-101310 47C | VersaWeigh - Power-User Option Pack |
PID-10131047B also known as PID-101310 47B | VersaWeigh - Electrical Option Pack |
PID-10131047A also known as PID-101310 47A | VersaWeigh - IP-66 Option Pack |
Maximize profits and minimize product giveaway with the Thermo Scientific™ Global VersaWeigh™ Checkweigher. This versatile, scalable system is still easy-to-use and maintain but now features more than 10 engineering and functionality improvements including frames designed to reduce food buildup, rapid-release, adjustable conveyors and belts, and high-torque sealed brushless motors for trouble-free operation. The fixed-height Global VersaWeigh is ideal for products up to 10 kilograms (22 pounds) and performs with an accuracy equal to or better than market requirement standards.
The Global VersaWeigh Checkweigher Features:
Compare and find the checkweigher that’s right for you.
View checkweighers ›
For pet food products in cartons, cans, bottles, or pouches, a chain checkweigher offers line speeds up to 700 packages per minute. It is appropriate for both dry and wet environments.
To ensure that pet food processors are not under or overfilling packages, checkweighers are used to verify the correct weight of each package. Accurate weight avoids giving away costly product or from receiving hefty fines from regulatory bodies for underfilling.
Checkweighers do not detect physical contaminants but they can be combined with food metal detectors and X-ray inspection systems that do detect contaminants. Food metal detection systems provide reliable, cost-effective protection from even the smallest metal contaminants found anywhere in a food production process. Food X-ray inspection systems provide protection from metal, glass, stone and other dense foreign objects for almost any type of packaged, bulk, or piped product.
Food manufacturers can find metallic and non-metallic foreign objects and eliminate “wet” product effects common with metal detectors with the Thermo Scientific NextGuard X-ray Detection Systems (https://www.thermofisher.com/order/catalog/product/NEXTGUARDPRO). Designed for a wide variety of food applications, NextGuard systems offer enhanced capabilities to inspect packaged products for missing pieces or components, under and over-fills, and other quality problems.
Checkweighers can sometimes come in contact with food products that can leak or spill. The equipment has to get washed down regularly to prevent contamination so it must meet IP65 Washdown requirements. Therefore, checkweigher components require materials that can withstand washdown using chemicals. The metal should be fully constructed of ANSI Type 304 SS (stainless steel), which is an austenite steel. For even harsher situations, ANSI 316 stainless is used; it is more impervious to corrosion than 304 SS. Equipment should meet American National Standard ANSI/IEC 60529-2004 for equipment with electrical components. This standard describes a system for classifying the degrees of protection provided by enclosures of electrical equipment for two conditions:
- The protection of persons against access to hazardous parts and protection of equipment against the ingress of solid foreign objects
- The ingress of water
Equipment should be designed with minimal flat, horizontal surfaces and slots to reduce food buildup, and use stainless steel conveyor beds. IP65 enclosures provide basic waterproof protection from water, including jet sprays used during washdown.
Proper design of the in-line checkweigher can influence accuracy. Here are some of the design factors:
- Product speed
- Product weight and size
- Product transfer onto and off of the checkweigher
- Reject method and design
In addition, conveyors must be level and aligned, belts and chains should be inspected regularly for wear or product buildup, gaps between conveyors should be minimized and infeed and weigh table belts or chains should be running at the same speeds. Other factors include the spacing between products' centers to ensure that more than one product is not on the weighing table at any one time, and calibrations should be done regularly.
Share catalog number, name or link