IPlus! Measurement and Control System for Nonwovens
IPlus! Measurement and Control System for Nonwovens
Thermo Scientific™
IPlus! Measurement and Control System for Nonwovens
Produce high performance nonwovens with a measurement and control system optimized for spunbond, spunlace, airlaid, carded, and meltblown processes.
Have Questions?
Catalog number PID10148820
Price (USD)/ Each
-
Request A Quote
Configuration:
L400 scanner, CLS Beta sensor, LineSpeed control
Nonwoven manufacturing processes must be carefully controlled to ensure that the fabrics are resilient, liquid repellant, and leakproof. Now you can overcome process challenges and produce high performance nonwoven fabric products with the Thermo Scientific™ IPlus! Measurement and Control System for Nonwovens. The IPlus! online system is optimized for the spunbond, spunlace, carded, airlaid, and needlepunch processes to control raw material distribution across the web. Closed loop Machine Direction control can also be added to enhance quality.
The IPlus! Measurement and Control System combines advanced sensor technology and powerful software in an intuitive, easy-to-use system to provide high performance and a low cost of ownership.
Features:
  • Basis weight, direct thickness, and or moisture weight measurement.
  • High resolution sensor provides accurate, real-time measurements.
  • Reduce scrap with precise process control.
  • Easier data management and archiving with the user inerface.
  • High reliability for less downtime.
  • Up to 2000 measurement profile points.
  • Advanced application controls.
  • Intuitive WonderWare HMI operator displays.
  • Remote support capability and advanced system diagnostics.
  • System software backup for quick and easy system restoration in case of accidental damage.
Nonwovens Package:
Thermo Scientific™ L400 Scanner
Thermo Scientific™ Prosis™ Infrared Thickness Sensor for total weight/thickness and moisture content measurement or Thermo Scientific™ Beta Plus™ Sensor for total weight measurement
LineSpeed Control
Specifications
ApplicationsNon-woven Materials
DescriptionIPlus! for Nonwovens
ConfigurationL400 scanner, CLS Beta sensor, LineSpeed control
Unit SizeEach