PowerEase™ Touch 350W Power Supply and SureLock™ Tandem Midi PVDF Welcome Pack
PowerEase™ Touch 350W Power Supply and SureLock™ Tandem Midi PVDF Welcome Pack
Invitrogen™

PowerEase™ Touch 350W Power Supply and SureLock™ Tandem Midi PVDF Welcome Pack

The PowerEase Touch 350W Power Supply* and SureLock Tandem Midi PVDF Welcome Pack contains: • PowerEase Touch 350W Power SupplyRead more
Have Questions?
Catalog number PSC350STMPV
Price (USD)/ 1 pack
2,904.65
Online exclusive
2960.00 
Save 55.35 (2%)
-
Add to cart
Price (USD)/ 1 pack
2,904.65
Online exclusive
2960.00 
Save 55.35 (2%)
Add to cart
PowerEase™ Touch 350W Power Supply and SureLock™ Tandem Midi PVDF Welcome Pack
Catalog numberPSC350STMPV
Price (USD)/ 1 pack
2,904.65
Online exclusive
2960.00 
Save 55.35 (2%)
-
Add to cart
The PowerEase Touch 350W Power Supply* and SureLock Tandem Midi PVDF Welcome Pack contains:

PowerEase Touch 350W Power Supply (115 VAC)* or PowerEase Touch 350W Power Supply (230 VAC)* or PowerEase Touch 350W Power Supply (230 VAC) (Australia)* or PowerEase Touch 350W Power Supply (230 VAC) (China)*
SureLock Tandem Midi Gel Tank
• Two SureLock Tandem Midi Blot Modules
SureLock Tandem Transfer Tray
SureLock Tandem Midi Pre-cut Membranes and Filter, 0.45 μm, PVDF

*dependent on region

About the PowerEase Touch 350W Power Supply
The Invitrogen PowerEase Touch 350W Power Supply is a high-throughput power supply with a 4.3-inch backlit LCD touchscreen. The touchscreen and user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for high current applications like high throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE, and western blotting. The PowerEase Touch 350W Power Supply offers four sets of output jacks that can be used simultaneously and three modes: constant voltage, constant current, and constant power for flexibility of use and efficiency.

About the SureLock Tandem Midi PVDF Welcome Pack
The SureLock Tandem Midi Welcome Pack, PVDF, is designed specifically for electrophoresis and wet transfer of Invitrogen pre-cast midi gels using PVDF transfer membranes. The innovative 2-in-1 gel electrophoresis and transfer tank enables electrophoresis and transfer of high-performance Invitrogen midi gels using the same tank.
Specifications
For Use With (Application)Gel Electrophoresis
Gel SizeMidi
Product LinePowerEase
Quantity1 pack
Shipping ConditionRoom Temperature
Unit Size1 pack