Sludge and Suspended Matter Testing Products
Sludge and suspend matter testing products are excellent for the determination of solid content, viscosity, and settleability properties in liquid samples. Testing products include settlometer kits, settling cones, and cone racks. Thermo Scientific™ Nalgene™ Settlometer Kit, an all-plastic tool for wastewater treatment plant operators, is used for analyzing sludge quality and aid in analyzing the activated sludge process.