MAS™ PAR™ TDM Controls
MAS™ PAR™ TDM Controls
Actual product may vary
Thermo Scientific™
MAS™ PAR™ TDM Controls
Check therapeutic drug monitoring test procedures with ready-to-use, liquid assayed multi-analyte MAS PAR TDM Controls.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLevel
PTD1-10011
PTD2-20022
PTD3-30033
Catalog number PTD1-1001
Price (USD)
-
Request A Quote
Level:
1
Check therapeutic drug monitoring test procedures with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ PAR™ TDM Controls, therapeutic drug controls with assayed values designed for therapeutic drugs and common immunoassayed test procedures on most automated instruments.
  • Liquid, ready to use
  • Features three significant levels
  • Includes values for major automated analyzers
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionMAS PAR TDM Quality Control
Level1
Quantity6 x 5 mL
Stability36 months
TypeControl
Detectable AnalytesAcetaminophen, Amikacin, Caffeine, Carbamazepine, Digoxin, Disopyramide, Estriol, Ethanol, Ethosuximide, Gentamicin, Lidocaine, Lithium, Methotrexate, N-acetylprocainamide (NAPA), Phenobarbital, Phenytoin, Primidone, Procainamide, Quinidine, Salicylate (SALY), Theophylline, Thyroid Stimulating Hormone (TSH), Thyroxine (T4 Total), Tobramycin, Tricyclic Antidepressants, Triiodthyronine Total (TT3), Valproic Acid, Vancomycin
Sample TypeSerum
Unit SizeEach
Contents & Storage
2°C to 8°C
lab-link
LabLink xL quality assurance software
Real-time, web-based data processing, peer data comparison, reports and documentation.
View MAS QC package inserts
Log in to LabLink xL
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection