Thermo Scientific™

LanthaScreen™ Tb-labeled Anti-Goat Antibody Kit

Catalog number: PV3769
Have Questions?
Thermo Scientific™

LanthaScreen™ Tb-labeled Anti-Goat Antibody Kit

Catalog number: PV3769
Have Questions?
Catalog Number
PV3769
Unit Size
25 µg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV376925 µg
-
PV37711 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
To facilitate in-house assay development around multiple target classes and assay formats, Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents. Tb-labeled secondary antibodies bind to their corresponding species IgG with low nanomolar apparent Kd's, and show negligible cross reactivity with IgG's from other species. Each Tb-labeled anti-species antibody is provided with a corresponding fluorescein-labeled IgG as a positive control.

In a kinase assay utilizing the Tb-labeled antibody generic reagent, anti-phosphopeptide primary antibody is not labeled and the detection step is performed by the addition of the Tb-labeled secondary antibody. This approach can be applied to other target classes and assay formats.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
25 μg
Reactivity
Goat
Shipping Condition
Dry Ice
Conjugate
Lanthanide Chelates, Terbium (Tb)
Product Line
LanthaScreen™

Contents & Storage

Store in freezer (-5 to -30°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text