Invitrogen™

LanthaScreen™ Tb-labeled Anti-Rabbit Antibody Kit

Catalog number: PV3775
Have Questions?
Invitrogen™

LanthaScreen™ Tb-labeled Anti-Rabbit Antibody Kit

Catalog number: PV3775
Have Questions?
Catalog Number
PV3775
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PV37751 mg
-
PV377325 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

To facilitate in-house assay development around multiple target classes and assay formats, Tb-labeled species-specific secondary antibodies are available within the LanthaScreen™ Toolbox of assay reagents. Tb-labeled secondary antibodies bind to their corresponding species IgG with low nanomolar apparent Kd's, and show negligible cross reactivity with IgG's from other species. Each Tb-labeled anti-species antibody is provided with a corresponding fluorescein-labeled IgG as a positive control.

In a kinase assay utilizing the Tb-labeled antibody generic reagent, anti-phosphopeptide primary antibody is not labeled and the detection step is performed by the addition of the Tb-labeled secondary antibody. This approach can be applied to other target classes and assay formats.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
1 mg
Reactivity
Rabbit
Shipping Condition
Dry Ice
Conjugate
Lanthanide Chelates, Terbium (Tb)
Product Line
LanthaScreen™

Contents & Storage

Store in freezer (-5°C to -30°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text