Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 µg, 10 µg and 20 µg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 µL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase.
Protein Form: Recombinant, Full Length
STK24 (MST3) is a serine/threonine kinase that is a member of the GCK subfamily of STE20-like proteins. The yeast 'Sterile 20' gene (STE20) functions upstream of the mitogen-activated protein kinase (MAPK) cascade. In mammals, protein kinases related to STE20 can be divided into 2 subfamilies based on their structure and regulation. Members of the PAK subfamily (see PAK3; MIM 300142) contain a C-terminal catalytic domain and an N-terminal regulatory domain that has a CDC42 (MIM 116952)-binding domain. In contrast, members of the GCK subfamily (see MAP4K2; MIM 603166), also called the Sps1 subfamily, have an N-terminal catalytic domain and a C-terminal regulatory domain without a CDC42-binding domain. STK24 belongs to the GCK subfamily of STE20-like kinases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 95; Mammalian STE20-like protein kinase 3; MST-3; OTTHUMP00000018594; RP11-111L24.5; serine/threonine kinase 24 (STE20 homolog, yeast); Serine/threonine-protein kinase 24; STE20-like kinase 3; STE20-like kinase MST3; sterile 20-like kinase 3
Gene Aliases: HEL-S-95; MST3; MST3B; STE20; STK24; STK3
UniProt ID: (Human) Q9Y6E0
Entrez Gene ID: (Human) 8428
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support