LanthaScreen™ TR-FRET RXR beta Coactivator Assay Kit, goat
Product Image
Thermo Scientific™

LanthaScreen™ TR-FRET RXR beta Coactivator Assay Kit, goat

This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15120:The LanthaScreen™ TR-FRET Retinoid X ReceptorRead more
Have Questions?
Catalog number PV4385
Price (USD)/ 800 x 20 µL assays
2,155.00
-
Add to cart
Price (USD)/ 800 x 20 µL assays
2,155.00
Add to cart
LanthaScreen™ TR-FRET RXR beta Coactivator Assay Kit, goat
Catalog numberPV4385
Price (USD)/ 800 x 20 µL assays
2,155.00
-
Add to cart
This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15120:

The LanthaScreen™ TR-FRET Retinoid X Receptor (RXR) beta Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential RXR beta ligands as agonists of coactivator recruitment. The homogeneous mix-and-read assay format results in a ligand EC50 composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (Figure 1).

How it works

The LanthaScreen™ TR-FRET RXR beta Coactivator Assay Kit includes a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and a human RXR beta ligand-binding domain (RXR beta-LBD) that is tagged with glutathione-S-transferase (GST). To assay, the RXR beta-LBD is added to ligand test compounds, followed by addition of a mixture of the fluorescein-coactivator peptide and Tb-anti-GST antibody. After room temperature incubation, the TR-FRET 520:495 emission ratio is calculated and used to determine the EC50 from a dose response curve of the compound

Contents and Storage:

The LanthaScreen™ TR-FRET RXR beta Coactivator Assay Kit contains RXR beta-LBD (GST) protein, fluorescein-labeled D22 coactivator peptide, Tb-anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateTb (Terbium)
Detection MethodFluorescence
For Use With (Equipment)Microplate Reader
Quantity800 x 20 μL Assays
Packaging384-well Plate
Assay EntryBiochemical coregulator interaction
Gene ID (Entrez)6257
ReadoutEnd Point
Shipping ConditionDry Ice
Target EntryRXR beta, NR2B2, RXRB
Product LineLanthaScreen™
For Use With (Application)Co-Factor Interaction Assay, TR-FRET
LigandRXR beta
Product TypeTR-FRET RXR Beta Coactivator Assay Kit
Unit Size800 x 20 µL assays
Contents & Storage
Store in ultra-cold freezer (-68 to -85°C).