A truncated version (residues 1-179) of c-Jun has been expressed as a fusion with GFP (Green Fluorescent Protein) to actRead more
Have Questions?
Catalog number PV4446
Price (USD)/ 20 nmol
1,062.00
-
Add to cart
Price (USD)/ 20 nmol
1,062.00
Add to cart
GFP-C-Jun (1-179)
Catalog numberPV4446
Price (USD)/ 20 nmol
1,062.00
-
Add to cart
A truncated version (residues 1-179) of c-Jun has been expressed as a fusion with GFP (Green Fluorescent Protein) to act as a substrate for the JNK family of kinases.
For use in combination with the LanthaScreen™ Tb-anti-c-Jun [pSer73] antibody (PV4453) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.