LanthaScreen™ TR-FRET TR beta Coactivator Assay Kit, goat
Product Image
Thermo Scientific™

LanthaScreen™ TR-FRET TR beta Coactivator Assay Kit, goat

This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15135:The LanthaScreen™ TR-FRET TR (thyroid receptor)Read more
Have Questions?
Catalog number PV4686
Price (USD)/ 800 x 20 µL assays
2,105.00
-
Add to cart
Price (USD)/ 800 x 20 µL assays
2,105.00
Add to cart
LanthaScreen™ TR-FRET TR beta Coactivator Assay Kit, goat
Catalog numberPV4686
Price (USD)/ 800 x 20 µL assays
2,105.00
-
Add to cart

This kit contains Goat Tb-Anti-GST antibody; other kit components are the same as kit A15135:

The LanthaScreen™ TR-FRET TR (thyroid receptor) beta Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential TR beta ligands as agonists of ligand-dependent coactivator recruitment. The kit uses a terbium (Tb)-labeled anti-GST antibody, a fluorescein-labeled coactivator peptide, and a human recombinant TR beta ligand-binding domain (TR beta-LBD) that is tagged with glutathione-S-transferase (GST) in a homogeneous, mix-and-read assay format.

To Assay:

When performing the LanthaScreen™ TR-FRET TR beta coactivator assay, TR beta-LBD is added to ligand test compounds followed by addition of a mixture of the fluorescein-coactivator peptide and Tb anti-GST antibody. After an incubation period at room temperature, the 520 nm/495 nm TR-FRET ratio is calculated and can be used to determine the EC50 from a dose response curve of the compound. Based on the biology of the TR beta-coactivator peptide interaction, the ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, induce a conformational change, and recruit coactivator peptide (Figure 1).

Contents and Storage:

The LanthaScreen™ TR-FRET TR beta Coactivator Assay Kit contains TR beta-LBD (GST) protein, fluorescently labeled SRC2-2 coactivator peptide, Tb-labeled anti-GST antibody, and buffers. Store components as indicated in the assay protocol (-80°C, -20°C, or +4°C).

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateTb (Terbium)
Detection MethodFluorescence
For Use With (Equipment)Microplate Reader
Packaging384-well Plate
Assay EntryBiochemical coregulator interaction
ReadoutEnd Point
Shipping ConditionDry Ice
Target EntryT3RB, TR beta, THRB, NR1A2
Product LineLanthaScreen™
For Use With (Application)Co-Factor Interaction Assay, TR-FRET
LigandTR Beta
Product TypeTR-FRET TR Beta Coactivator Assay Kit
No. of Assays800 x 20 μL Assays
Unit Size800 x 20 µL assays
Contents & Storage
Store in ultra-cold freezer (-68°C to -85°C).