LanthaScreen™ Eu-anti-GST Antibody
LanthaScreen™ Eu-anti-GST Antibody

LanthaScreen™ Eu-anti-GST Antibody

LanthaScreen™ europium-labeled anti-GST goat antibodies are used to indirectly label proteins tagged with GST. This can be advantageous when workingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
PV55951 mg
PV559425 μg
2 Options
Catalog number PV5595
Price (USD)/ 1 mg
12,087.65
Online exclusive
13450.00 
Save 1,362.35 (10%)
-
Add to cart
Quantity:
1 mg
Price (USD)/ 1 mg
12,087.65
Online exclusive
13450.00 
Save 1,362.35 (10%)
Add to cart
LanthaScreen™ Eu-anti-GST Antibody
Catalog numberPV5595
Price (USD)/ 1 mg
12,087.65
Online exclusive
13450.00 
Save 1,362.35 (10%)
-
Add to cart
LanthaScreen™ europium-labeled anti-GST goat antibodies are used to indirectly label proteins tagged with GST. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Eu chelates negatively impacts protein activity. These antibodies can be used with our Kinase Tracers to conduct LanthaScreen™ Eu Kinase Binding Assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AntigenGST
ApplicationsKinase Assay, Kinase Assay
Concentration0.25 mg/mL, 0.25 mg/ml, 0.25 mg⁄ml
ConjugateEuropium
Molecular Weight of Antigen150kDa
Quantity1 mg
Regulatory StatusRUO, RUO
Research DisciplinePharma and Biopharma
Primary or SecondaryPrimary
Packaging1 Kit(s)
Antibody Molecular Weight150 kDa
FormLiquid
Excitation/Emission340⁄665
For Use With (Equipment)Microplate Reader
Shipping ConditionDry Ice
Target MoleculeGST
TechniqueTR-FRET
Product LineLanthaScreen™
Unit Size1 mg
Contents & Storage
Europium-labeled anti-GST antibody is supplied in HEPES-buffered saline. Store at -20°C.