Matched and validated lipid and peptide substrates are available to complement the Adapta® Universal Kinase Assay. The assays developed withRead more
Have Questions?
Catalog Number
Quantity
PV5707
1 mL
Catalog number PV5707
Price (USD)
1,178.00
Each
Estimated availability date 25-Aug-2025
Add to cart
Quantity:
1 mL
Recurring order eligible. Learn more »
Price (USD)
1,178.00
Each
Add to cart
Ask our AI about this Product
Matched and validated lipid and peptide substrates are available to complement the Adapta® Universal Kinase Assay. The assays developed with these reagents allow for primary or secondary screening of kinase inhibitors.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Amino Acid SequenceARTKQTARKSTGGKAPRKQLW
Assay EntryBiochemical ADP accumulation
Quantity1 mL
ReadoutEnd Point
Shipping ConditionDry Ice
SubstratePeptide based on the N-terminus of Histone H3, Peptide substrate based on the N-terminus of Histone H3
Target EntryGSG2
Validated KinasesGSG2 (haspin)
Detection MethodFluorescence
FormLiquid
Substrate PropertiesPeptide-Based Substrate
Substrate TypeKinase Substrate
Target EnzymeGSG2 (haspin)
Unit SizeEach
Contents & Storage
2 mM Histone H3 (1-20) peptide substrate in 50 mM HEPES (pH 7.5), 0.01% Brij-35, 10 mM MgCl2, and 1 mM EGTA. Store at -20°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.