Thermo Scientific™

Pre-Weigh Buffered Peptone Water (BPW) Agar

Catalog number: PW0509-8L
Have Questions?
Thermo Scientific™

Pre-Weigh Buffered Peptone Water (BPW) Agar

Catalog number: PW0509-8L
Have Questions?
Speed up your workflow when preparing microbial culture medium for pre-enrichment of Salmonella species from foods with Thermo Scientific™ Pre-weigh Buffered Peptone Water. The pre-weighed sachets of Thermo Scientific™ Oxoid™ dehydrated culture media are designed for single use when preparing 1 L or 8 L volumes of your medium. Save valuable time and start testing quickly by removing the requirement to weigh powder—just rip, tip and pour.
 
Catalog Number
PW0509-8L
Unit Size
Each
Quantity
10 x Sachets for 8 L Medium
Price (USD)
Full specifications
DescriptionPre-Weigh Buffered Peptone Water (BPW)
FormPowder
Product TypeAgar
Quantity10 x Sachets for 8 L Medium
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
20 x Sachets for 1 L Medium
Price (USD)
Full specifications
DescriptionPre-Weigh Buffered Peptone Water (BPW)
FormPowder
Product TypeAgar
Quantity20 x Sachets for 1 L Medium
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
PW0509-8LFull specifications
Each10 x Sachets for 8 L MediumRequest A Quote
DescriptionPre-Weigh Buffered Peptone Water (BPW)
FormPowder
Product TypeAgar
Quantity10 x Sachets for 8 L Medium
Unit SizeEach
PW0509-1LFull specifications
Each20 x Sachets for 1 L MediumRequest A Quote
DescriptionPre-Weigh Buffered Peptone Water (BPW)
FormPowder
Product TypeAgar
Quantity20 x Sachets for 1 L Medium
Unit SizeEach
Showing 2 of 2
Pre-Weigh Buffered Peptone Water sachets contain Thermo Scientific Oxoid Buffered Peptone Water medium (Manf. Part No. CM0509). They are part of a range of our most frequently used and well-characterized dehydrated culture media, which is now available in handy pre-weighed foil packages.


Simply tear open the selected media sachet, add to the required volume of water, mix, and autoclave as usual.


  • Efficient: choose from a full range of pre-weighed media formats for reduced hands-on prep time and maximum productivity
  • Convenient: ‘rip and tip’ sachets enable faster media production and simplified workflows
  • Compliant: proven and validated media formats deliver confidence and reliable performance
  • Productive: reduce routine dehydrated culture media handling and weighing to spend more time on value-added tasks
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Laboratory Use.
For Laboratory Use

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text