Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits
Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits
Actual product may vary
Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits
Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits
Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits
Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits
Invitrogen™

Qubit™ RNA High Sensitivity (HS), Broad Range (BR), and Extended Range (XR) Assay Kits

Kits enable quick and sensitive detection of low and high abundance RNA, and can distinguish between RNA and DNA, protein, and free nucleotides.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityAssay
Q10210100 assaysRNA Quantification, BR
Q10211500 assaysRNA Quantification, BR
Q33223100 assaysRNA Quantification, ext range
Q33224500 assaysRNA Quantification, ext range
Q32852100 assaysRNA Quantification, HS
Q32855500 assaysRNA Quantification, HS
Catalog number Q10210
Price (EUR)
135,65
Online Exclusive
160,00
Save 24,35 (15%)
Each
-
Add to cart
Quantity:
100 assays
Assay:
RNA Quantification, BR
Price (EUR)
135,65
Online Exclusive
160,00
Save 24,35 (15%)
Each
Add to cart
Ask our AI about this Product
Achieve accurate and precise quantification of RNA with Qubit RNA HS, BR, and XR Assay Kits. These RNA quantification kits enable quick and selective detection of low and high abundance RNA samples, and can distinguish RNA from DNA, protein, and free nucleotides. Contaminants such as salts, solvents, or detergents are well-tolerated.
Qubit RNA HS, BR, and XR Assay Kits, designed for use with Qubit Fluorometers, are highly selective for RNA over DNA, protein, and free nucleotides. All kits provide concentrated assay reagent, dilution buffer, and pre-diluted RNA standards. Simply dilute the reagent using the buffer provided, add your sample (any volume between 1 μL and 20 μL is acceptable), and read the concentration using a Qubit Fluorometer.

Qubit RNA HS Assay Kit
The Qubit RNA HS (High Sensitivity) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of low-abundance RNA samples.  Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 0.2 to 200 ng/μL, providing a detection range of 4−200 ng.

Qubit RNA BR Assay Kit
The Qubit RNA BR (Broad-Range) Assay Kit, when used with the Qubit Fluorometer, provides an accurate and selective method for the quantitation of RNA samples.  Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 0.5 to 1,200 ng/μL, providing a detection range of 10−1,200 ng.

Qubit RNA XR Assay Kit

The Qubit RNA XR (Extended Range) Assay Kit, designed for use with Qubit 4 and Qubit Flex Fluorometers, provides an accurate and selective method for the quantitation of high-abundance RNA samples. Depending on sample volume, the assay kit is designed to be accurate for initial RNA sample concentrations of 5 to 20,000 ng/μL, providing a detection range of 100−20,000 ng.

Notes
• Qubit RNA HS and BR Assay kits can be used with any Qubit Fluorometer
• Qubit RNA XR Assay Kits are designed to be used only with Qubit 4 or Qubit Flex Fluorometers
• Use with thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 Fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex Fluorometer

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayRNA Quantification, BR
Excitation/Emission644/673
For Use With (Equipment)Qubit Fluorometer
No. of Reactions100 Reactions
Product LineQuant-iT
Quantitation Range10 to 1200 ng
Quantity100 assays
Shipping ConditionRoom Temperature
Detection MethodFluorescence
Unit SizeEach

Enhance your Qubit Fluorometer with the latest firmware

New Qubit 4 and Qubit Flex firmware features include importing sample IDs, renaming files before exporting, a countdown timer, and on-board troubleshooting guidance. Get the most out of your Qubit assays with these powerful new tools.

Download the latest Qubit firmware ›

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link