Qdot™ 565 ITK™ Amino (PEG) Quantum Dots
Qdot™ 565 ITK™ Amino (PEG) Quantum Dots
Invitrogen™

Qdot™ 565 ITK™ Amino (PEG) Quantum Dots

Qdot™ 565 ITK™ amino (PEG) quantum dots are the ideal starting material for preparing custom conjugates of ultrabright and photostableRead more
Have Questions?
Catalog number Q21531MP
Price (USD)/ 250 µL
850.00
-
Add to cart
Price (USD)/ 250 µL
850.00
Add to cart
Qdot™ 565 ITK™ Amino (PEG) Quantum Dots
Catalog numberQ21531MP
Price (USD)/ 250 µL
850.00
-
Add to cart
Qdot™ 565 ITK™ amino (PEG) quantum dots are the ideal starting material for preparing custom conjugates of ultrabright and photostable fluorescently labeled proteins or other biopolymers. These probes are functionalized with amine-derivatized PEG, which prevents non-specific interactions and provides a convenient handle for conjugation. The amino quantum dots react efficiently with isothiocyanates and succinimidyl esters, or with native carboxylic acids using water-soluble carbodiimides such as EDC. Such derivatives may be used for various labeling and tracking applications that require ultrabright and stable fluorescence. Our Qdot™ ITK™ amino quantum dots are provided as 8 μM solutions and are available in 8 colors of Qdot™ probes.

Important Features of Qdot™ ITK™ Amino Quantum Dots:
• Qdot™ 565 ITK™ amino quantum dot has emission maxima of ∼565 nm
• Extremely photostable and bright fluorescence
• Efficiently excited with single-line excitation sources
• Narrow emission, large stokes shift
• Available in multiple colors
• Ideal for various labeling and tracking applications


Properties of Qdot™ Nanocrystals
Qdot™ probes are ideal for imaging and labeling applications that require bright fluorescent signals and/or real-time tracking. Unique among fluorescent reagents, all nine available colors of Qdot™ probes can be simultaneously excited with a single (UV to blue-green) light source. This property makes these reagents excellent for economical and user-friendly multiplexing applications. Qdot™ labels are based on semiconductor nanotechnology and are similar in scale to moderately sized proteins.

About the Innovator’s Tool Kit Qdot™ ITK™ Reagents
These Qdot™ ITK™ probes are ideal for researchers who wish to prepare specific (non-stocked) conjugates for their applications and need customizable conjugation functionality.

Other Forms of Qdot™ Nanocrystals are Available
In addition to the amine-derivatized form, we offer Qdot™ ITK™ quantum dots with carboxyl and aliphatic hydrocarbon modifications. We’ve also developed a wide range of Qdot™ nanocrystals conjugates and labeling kits. Investigate the properties of Qdot™ nanocrystals or read the Molecular Probes™ Handbook Section 6.6—Qdot™ Nanocrystals to find out more.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration8 μM
Quantity250 μL
Product TypeQuantum Dot
Chemical ReactivityCarboxylic Acid, Ketone, Aldehyde
Shipping ConditionRoom Temperature
Reactive MoietyAmine, Primary Amine
Label or DyeQdot™ 565
Label TypeQdot Nanocrystals
Product LineITK™, Qdot™
Emission565
Unit Size250 µL
Contents & Storage
Store in refrigerator (2–8°C).