Yeast Nitrogen Base
Product Image
Invitrogen™

Yeast Nitrogen Base

Yeast nitrogen base (YNB) is a base medium for preparation of minimal and synthetic defined yeast media. YNB contains ammoniumRead more
Have Questions?
Catalog NumberQuantity
Q300071 Pouch
Catalog number Q30007
Price (USD)
268.00
Each
-
Add to cart
Quantity:
1 Pouch
Price (USD)
268.00
Each
Add to cart
Ask our AI about this Product
Yeast nitrogen base (YNB) is a base medium for preparation of minimal and synthetic defined yeast media. YNB contains ammonium sulfate but does not contain amino acids. Both bulk and convenient single-use pouches are available.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 Pouch
FormPowder
Product TypeMicrobiology Media
Target Organism ClassP. pastoris, S. cerevisiae
Unit SizeEach
Contents & Storage
Each pouch contains 67 g of reagent to prepare 500 mL of 10X YNB for Pichia media or 1,000 mL of 10X YNB for S. cerevisiae media.

Store at room temperature. Guaranteed stable for 6 months when properly stored.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

What are the components and concentrations for 1X YNB (yeast nitrogen base)?

For the pouches sold as Cat. No. Q30007, each pouch contains reagents to prepare 500 mL of 10X YNB for Pichia media or 1,000 mL of 10X YNB for S. cerevisiae media (since Pichia media is 2X what is used for S. cerevisiae). Below is the formulation for 1X YNB:

Ammonium sulfate, 5 g
Biotin, 0.002 mg
Calcium pantothenate, 0.4 mg
Folic acid, 0.002 mg
Inositol, 2 mg
Niacin, 0.4 mg
Para-aminobenzoic acid, 0.2 mg
Pyridoxine HCl, 0.4 mg
Riboflavin, 0.2 mg
Thiamine HCl, 0.4 mg
Boric acid, 0.5 mg
Copper sulfate, 0.4 mg
Potassium iodide, 0.1 mg
Ferric chloride.6H2O, 0.2 mg
Manganese sulfate.H2O, 0.4 mg
Sodium molybdate.2H2O, 0.2 mg
Zinc sulfate.H2O, 0.4 mg
Potassium phosphate monobasic, 1 g
Magnesium sulfate, 0.5 g
Sodium chloride, 0.1 g
Calcium chloride, 0.1 mg

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Can YNB medium (with ammonium sulfate, without amino acids) be autoclaved instead of filter-sterilized?

Yes, YNB medium with ammonium sulfate and without amino acids can be autoclaved for sterilization.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link