Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Detection Method | Fluorescence |
For Use With (Equipment) | Qubit Fluorometer |
For Use With (Application) | microRNA Quantitation |
No. of Reactions | 100 Reactions |
Product Line | Qubit |
Product Type | microRNA Assay Kit |
Shipping Condition | Room Temperature |
Excitation/Emission | 498/518 |
Unit Size | 100 assays |
Detection Method | Fluorescence |
For Use With (Equipment) | Qubit Fluorometer |
For Use With (Application) | microRNA Quantitation |
No. of Reactions | 500 Reactions |
Product Line | Qubit |
Product Type | microRNA Assay Kit |
Shipping Condition | Room Temperature |
Excitation/Emission | 498/518 |
Unit Size | 500 assays |
Catalog Number | Specifications | Unit Size | No. of Reactions | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Q32880 | 100 assays | 100 Reactions | 128.65 Online price 130.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q32881 | 500 assays | 500 Reactions | 378.65 Online price 395.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|