Invitrogen™

Quant-iT™ microRNA Assay Kit

Catalog number: Q32882
Have Questions?
Invitrogen™

Quant-iT™ microRNA Assay Kit

Catalog number: Q32882
Have Questions?
Catalog Number
Q32882
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
Q328821 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Quantitate microRNA easily and accurately with the Quant-iT microRNA Assay Kit, a microRNA (miRNA) detection kit that detects miRNA even in the presence of common contaminants such as salts, free nucleotides, solvents, detergents, and protein. The assay is highly selective for microRNA over rRNA or large mRNA (>1000 bp).

Although not exclusively selective for miRNA, the assay reagent can reproducibly quantify miRNA in pure samples down to levels as low as 0.5 ng following the supplied protocol, even in the presence of mRNA.

Features of the Quant-iT microRNA Assay Kit include:
• Sensitive detection of as little as 0.5 ng miRNA in a microplate well.
• A core dynamic range of 5–500 ng/mL.
• Accurate readings of sample concentrations ranging from 50 ng/mL to 100 μg/mL.

The Quant-iT microRNA Assay Kit provides concentrated assay reagent, dilution buffer, and pre-diluted miRNA standards. Simply dilute the reagent 1:200, load 200 μL into the wells of a microplate, add 1–20 μL of sample, mix, and read the fluorescence. The assay is performed at room temperature, and the signal is stable for approximately three hours. Because the assay tolerates 1–20 μL of sample in an assay volume of 200 μL, accurate results can be obtained for initial sample concentrations from 50 ng/mL–100 μg/mL.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay
microRNA Quantitation
Detection Method
Fluorescence
Excitation/Emission
498/518
For Use With (Equipment)
Fluorometer, Microplate Reader
No. of Reactions
1000 (200 μL Assay Volume)
Quantitation Range
1 to 100 ng
Product Line
Quant-iT

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text