Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay | dsDNA Quantitation, high sensitivity |
Detection Method | Fluorescence |
For Use With (Equipment) | Microplate Reader |
No. of Reactions | 1000 Assays (200 μL assay volume) |
Quantitation Range | 0.2 to 100 ng |
Product Line | Quant-iT |
Shipping Condition | Room Temperature |
Unit Size | 1 kit |
Assay | dsDNA Quantitation, broad range |
Detection Method | Fluorescence |
For Use With (Equipment) | Microplate Reader |
No. of Reactions | 1000 Assays (200 μL assay volume) |
Quantitation Range | 4 to 1000 ng |
Product Line | Quant-iT |
Shipping Condition | Room Temperature |
Unit Size | 1 kit |
Catalog Number | Specifications | Unit Size | Assay | Quantitation Range | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Q33120 | 1 kit | dsDNA Quantitation, high sensitivity | 0.2 to 100 ng | 535.65 Online price 580.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q33130 | 1 kit | dsDNA Quantitation, broad range | 4 to 1000 ng | 536.65 Online price 584.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|