Qubit™ 4 Fluorometer, with WiFi
Invitrogen™

Qubit™ 4 Fluorometer, with WiFi

The Qubit 4 Fluorometer with WiFi is the latest version of the popular Qubit fluorometer designed to accurately measure DNA,Read more
Have Questions?
Catalog NumberQuantity
Q332381 fluorometer
Catalog number Q33238
Price (USD)
4,786.65
Online Exclusive
5,260.00
Save 473.35 (9%)
Each
Quantity:
1 fluorometer
Price (USD)
4,786.65
Online Exclusive
5,260.00
Save 473.35 (9%)
Each
The Qubit 4 Fluorometer with WiFi is the latest version of the popular Qubit fluorometer designed to accurately measure DNA, RNA, and protein quantity, and now also RNA integrity and quality, using the highly sensitive Qubit assays. The Qubit 4 Fluorometer was re-engineered to enable data transfer via WiFi as well as to run the Qubit RNA IQ (integrity & quality) assay. The Qubit 4 Fluorometer and RNA IQ Assay Kit work together to accurately distinguish intact from degraded RNA in just two easy steps.

For all Qubit assays, the concentration or quality of the target molecule in the sample is reported by a fluorescent dye that emits a signal only when bound to the target, which minimizes the effects of contaminants on the result. The easy-to-use touch-screen menus make it easy to select and run the assays you need, with results displayed in just a few seconds.

Key features of the Qubit 4 Fluorometer include:
• Fast and highly accurate quantitation of DNA, RNA, and protein in less than three seconds per sample
• Measures intact RNA in less than 5 seconds per sample
• High levels of accuracy using only 1–20 μL of sample, even with very dilute samples
• On-board Reagent Calculator quickly generates Qubit working solution preparation instructions
• Stores results from up to 1000 samples
• Large 5.7-inch, state-of-the-art color touch screen for easy workflow navigation
• Graphical display indicates when samples are in the extended range or out of range
• Small footprint saves space on your bench
• Flexibility to export data to using the included WiFi dongle, a USB drive, or directly to your computer via a USB cable
• Ability to personalize your Qubit fluorometer with the assays you run most, add new assays, or even create your own assays with the MyQubit software and web tool
• Language of your choice including English, French, Spanish, Italian, German, simplified Chinese, and Japanese
• Pre-loaded program to test performance of fluorometer using the Qubit Fluorometer Verification Assay Kit

Learn more about the Qubit Fluorometer ›

The Qubit 4 Fluorometer can also be used to directly measure the fluorescence of samples using Fluorometer mode. Additionally, Ion Sphere Particle quality can be evaluated on the Qubit 4 Fluorometer using the Ion Sphere Quality Control Kit prior to performing a sequencing run on the Ion PGM sequencer.

Enables more accurate results than with UV absorbance at low concentrations
The Qubit 4 Fluorometer is far more sensitive than UV absorbance, which measures anything absorbing at 260 nm, including DNA, RNA, protein, free nucleotides, or excess salts. Moreover, UV spectrophotometry is often not sensitive enough to accurately measure low concentrations of DNA and RNA. With the Qubit fluorometer, your research is enhanced by more accurate measurements because the dyes in the Qubit assay kits fluoresce only when bound to the selected molecule (DNA, RNA or protein) in your sample. This allows you to avoid repeating work due to inaccurate measurements.

Fast, simple, and accurate measurement of RNA quality
The Qubit RNA IQ assay provides a fast, simple method to check whether an RNA sample has degraded. The assay utilizes two unique dyes: one that binds to large, intact RNA, and another that selectively binds to small, degraded RNA. Together they enable you to quickly assess the quality and integrity of an RNA sample. To use, simply add your samples to the RNA IQ working solution, then measure on the Qubit 4 Fluorometer.

Save samples and bench space
The thoughtfully designed Qubit 4 Fluorometer requires samples of only 1–20 μL. It is intended for use at room temperature and occupies only a small amount of bench space. The Qubit 4 Fluorometer features advanced optics and data analysis algorithms, and comes equipped with a WiFi dongle, a USB thumb drive, and cable for data transfer to Excel™ software and for software downloads, as well as a universal power supply, four plug adapters, and electromagnetic CE certification. The Qubit 4 Fluorometer enables maximum flexibility for data export using WiFi, USB drive, or USB cable direct to computer. Additionally, all data can be saved in the Qubit cloud application, including direct transfer from the instrument via WiFi.

Single sample measurements are a breeze
This user-friendly fluorometer is combined with quick and simple assay procedures. Calculations and settings are automatically performed by the instrument. The Qubit assays for use with the Qubit 4 Fluorometer are all performed using the same general protocol, which uses a simple mix-and-read format with incubation times of only two minutes for the DNA and RNA assays. 500 L thin-walled PCR tubes (Cat. No. Q32856) are required.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)DNA Quantification, RNA Quantification, Protein Quantification
Quantity1 fluorometer
Sample Volume1 to 20 μL
Shipping ConditionRoom Temperature
Excitation Wavelength470 nm (Blue LED), 635 nm (Red LED)
For Use With (Equipment)Qubit Fluorometer
Product LineQubit
TypeFluorometer
Unit SizeEach

Frequently asked questions (FAQs)

What is the standard warranty of the Qubit 4 Fluorometer, with WiFi?

3 years is the standard warranty period.

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.

I would like to use my Qubit 4 Fluorometer with the Qubit 1X dsDNA BR Assay, but the assay is not listed as an option on the fluorometer. How can I resolve this?

The Qubit 4 Fluorometer needs to be updated. The Qubit 4 Fluorometer can be updated with the USB that was provided with the Qubit. If you do not have the USB, we recommend using an empty 2.0GB that is FAT32 formatted. If the USB is not recognized, leave USB plugged in for a few minutes before running the update. This should give you the assay you need to run the Qubit Assay.

You can find the updates on the Technical Resources for Qubit Fluorometers page.

My Qubit 4 Fluorometer, with Wifi (Cat. No. Q33238) has not installed the files to allow me to use the Qubit ssDNA Assay Kit. Could you provide the files I need?

To access the Qubit ssDNA Assay Kit on the Qubit 4, please try selecting the Oligo option from the home screen, and the ssDNA Assay should appear there. If not, please check if you need to update your instrument firmware to the latest version v2.19, which is available for download from the Technical Resources for Qubit Fluorometers page on thermofisher.com.
For standard/preloaded Qubit assays we do not provide individual .qbt files, but they are included in the firmware itself.

How can I use the Qubit Protein Broad Range Assay (Cat. No. A50668) on the Qubit 4 Fluorometer, with WiFi (Cat. No. Q33238)?

To be able to use the Qubit Protein Broad Range Assay (Cat. No. A50668) on the Qubit 4 Fluorometer (Cat. No Q33238) you would need to install the latest firmware on the fluorometer. Please find the latest Qubit 4 firmware (v2.19) on the following webpage: https://downloads.thermofisher.com/Qubit4/v2.19/Qubit4_update_v2.19.pak Save the firmware update on a USB stick and upload it to the Qubit 4 Fluorometer. Once the firmware update has been installed, you will be able to see the Protein BR assay on the Qubit 4 Fluorometer.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Quantitation Support Center.

How can I access user settings when using the Qubit 4 Fluorometer, with WiFi (Cat. No. Q33238)?

You have to create a user profile and log in. Please follow the instructions for login below:

  • Turn on the device
  • From the home screen, touch the person icon on the top left corner
  • Touch the “Get Started” button
  • Touch the “Create Profile” button
  • information for username and pin
  • Log in by touching the person icon and entering the user info in the sign in fields


Find additional tips, troubleshooting help, and resources within our Nucleic Acid Quantification Support Center.

Citations & References (56)

Citations & References
Abstract
Rapid sequencing of MRSA direct from clinical plates in a routine microbiology laboratory.
Authors:Blane B, Raven KE, Leek D, Brown N, Parkhill J, Peacock SJ,
Journal:J Antimicrob Chemother
PubMed ID:31039248
'Routine sequencing of MRSA could bring about significant improvements to outbreak detection and investigation. Sequencing is commonly performed using DNA extracted from a pure culture, but overcoming the delay associated with this step could reduce the time to infection control interventions. To develop and evaluate rapid sequencing of MRSA using ... More
A comparative analysis of drinking water employing metagenomics.
Authors:Brumfield KD, Hasan NA, Leddy MB, Cotruvo JA, Rashed SM, Colwell RR, Huq A
Journal:PLoS One
PubMed ID:32271799
'The microbiological content of drinking water traditionally is determined by employing culture-dependent methods that are unable to detect all microorganisms, especially those that are not culturable. High-throughput sequencing now makes it possible to determine the microbiome of drinking water. Thus, the natural microbiota of water and water distribution systems can ... More
Dietary Adaptation of Microbiota in Drosophila Requires NF-?B-Dependent Control of the Translational Regulator 4E-BP.
Authors:Vandehoef C, Molaei M, Karpac J,
Journal:Cell Rep
PubMed ID:32521261
'Dietary nutrients shape complex interactions between hosts and their commensal gut bacteria, further promoting flexibility in host-microbiota associations that can drive nutritional symbiosis. However, it remains less clear if diet-dependent host signaling mechanisms also influence these associations. Using Drosophila, we show here that nuclear factor ?B (NF-?B)/Relish, an innate immune ... More
Glucose-6-phosphate dehydrogenase correlates with tumor immune activity and programmed death ligand-1 expression in Merkel cell carcinoma.
Authors:Nakamura M, Nagase K, Yoshimitsu M, Magara T, Nojiri Y, Kato H, Kobayashi T, Teramoto Y, Yasuda M, Wada H, Ozawa T, Umemori Y, Ogata D, Morita A,
Journal:J Immunother Cancer
PubMed ID:33361404
'Merkel cell carcinoma (MCC) is a rare and highly malignant skin cancer. Some cases have a good prognosis and spontaneous regression can occur. Reported prognostic markers, such as Merkel cell polyoma virus infection or programmed death ligand-1 (PD-L1) expression, remain insufficient for precisely estimating the vastly different patient outcomes. We ... More
Geographical reconstruction of the SARS-CoV-2 outbreak in Lombardy (Italy) during the early phase.
Authors:Micheli V, Rimoldi SG, Romeri F, Comandatore F, Mancon A, Gigantiello A, Perini M, Mileto D, Pagani C, Lombardi A, Gismondo MR,
Journal:J Med Virol
PubMed ID:32816316
'The first identification of autochthonous transmission of SARS-CoV-2 in Italy was documented by the Laboratory of Clinical Microbiology, Virology and Bioemergencies of L. Sacco Hospital (Milano, Italy) on 20th February 2020 in a 38 years old male patient, who was found positive for pneumonia at the Codogno Hospital. Thereafter Lombardy ... More