The Qubit Fluorometer USB Flash Drive is designed for the transfer of data from the Qubit Fluorometer. Only the Qubit 2.0, 3, and 4 fluorometers have an available USB port. Data exported from the Qubit Fluorometer are stored in CSV format and can be opened by programs such as Microsoft Excel.
For Research Use Only. Not for use in diagnostic procedures.