Invitrogen™

QuantiGene™ Plex Automation Setup Kit

Catalog number: QGP-SETUP-384
Have Questions?
Invitrogen™

QuantiGene™ Plex Automation Setup Kit

Catalog number: QGP-SETUP-384
Have Questions?
Catalog Number
QGP-SETUP-384
Unit Size
6 tests
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
QGP-SETUP-3846 tests
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The QuantiGene Plex Automation Setup Kit enables setup, testing, and validation of instruments to automate crucial steps of the QuantiGene Plex 384-well assay protocol, such as liquid handling and plate washing, before running an actual assay with valuable sample. The user guide describes how to adjust and optimize an automated magnetic plate washer as needed during the QuantiGene Plex 384-well assay workflow in order to ensure optimal performance and avoid bead loss.

Each kit provides six optimization tests for automated magnetic plate washing.

Specifications

Format
384-well
For Use With (Equipment)
FLEXMAP 3D™ INTELLIFLEX™
Product Line
QuantiGene™ Plex
Product Type
Automation Setup Kit
Quantity
6 Tests
Shipping Condition
Wet Ice

Contents & Storage

• Automation Setup Beads (1X)
• SAPE Wash Buffer Concentrate (10X)
• QuantiGene Plex 384-well flat bottom magnetic separation plate
• Plate seals

Store at 2°C to 8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text