Search Thermo Fisher Scientific
Catalog Number | Voltage |
---|---|
QP0703 | 120 V |
QP0704 | 220 V |
VorTemp features:
• A microprocessor to control all parameters including temperature and timed operation
• Self-diagnostic software to continuously monitor for proper temperature and shaking speed, alerting the user to any errors
• Heating and shaking that are controlled by feedback loops which precisely maintain chosen parameters, independent of load or voltage fluctuations
• Sturdy construction for stability during high speed shaking
• An insulated chamber to maintain temperature and reduce operating noise
Share catalog number, name or link