Invitrogen™

QuantiGene™ Plex Assay Kit

Catalog number: QP1015
Have Questions?
Invitrogen™

QuantiGene™ Plex Assay Kit

Catalog number: QP1015
Have Questions?
Catalog Number
QP1015
Unit Size
10 plates
Price (USD)
Quantity
Catalog Number
Unit Size
1 plate
Price (USD)
Quantity
Catalog Number
Unit Size
3 plates
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
QP101510 plates
QP10131 plate
QP10143 plates
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The QuantiGene Plex Assay Kit contains the generic reagents, buffers, and plates necessary to perform the assay in 96-well plates, and must be used with a target-specific QuantiGene Plex panel (sold separately). See related sample preparation reagents and RNA controls .

The QuantiGene Plex Assay is a gene expression assay that enables multiplexed measurement of up to 80 RNA transcript targets. The assay uses target-specific probes to capture the RNA of interest, and branched DNA technology then amplifies fluorescent signals which are read with a Luminex xMAP instrument.

Difficult sample types—works with degraded and cross-linked RNA in FFPE tissues and blood
True multiplexing—measure up to 80 genes of interest and housekeeping genes in the same well with no cross-reactivity
Standardized platform—uses 96 or 384-well plate formats compatible with Luminex 200, MagPix, and FlexMap3D systems
Simple workflow—employs an ELISA-like workflow for direct hybridization of transcripts to beads and transcript labeling
Large inventory of validated genes—select from over 15,000 genes to create pathway and disease-themed panels
Rapid turnaround for custom panels—if we don’t have your gene(s), we can create a custom panel within two weeks

Technology
QuantiGene assays utilize branched DNA (bDNA) technology that has been used for decades in the VERSANT™ diagnostic assays for HIV-1, HCV, and HBV. First, cells are lysed or tissue samples are homogenized to release the target RNA. Second, the oligonucleotide probe set is incubated with the target RNA and Luminex xMAP bead set overnight. During this incubation, the probes cooperatively hybridize to the target and capture probes bound to each bead, capturing the target RNA. Third, signal amplification is performed via sequential hybridization of the bDNA preamplifier, amplifier, and label-probe molecules to the target. Addition of streptavidin phycoerythrin generates a luminescent signal directly proportional to the amount of target mRNA present in the sample.

Applications
QuantiGene Plex assay applications include compound screening of drug candidates, biomarker verification, siRNA knockdown efficiency, prospective and/or retrospective analysis of clinical samples, miRNA profiling, microarray validation, predictive toxicology, and detection of translocations and fusion genes.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
96-well, 96-well
For Use With (Application)
QuantiGene Plex Panel, QuantiGene Plex Panel
For Use With (Equipment)
Luminex instrument, Luminex instrument
Product Line
QuantiGene Plex, QuantiGene Plex
Product Type
Multiplex Assay Kit, Multiplex Assay Kit
Quantity
10 Plates, 10 Plates
Shipping Condition
Blue Ice, Blue Ice
Species
Universal, Universal
Sample Type
Purified RNA, cell/blood/FFPE lysates, tissue homogenates, Purified RNA, cell/blood/FFPE lysates, tissue homogenates
Detection Method
Fluorescence, Fluorescence
Unit Size
10 plates

Contents & Storage

• 350 μL Proteinase K, –20°C
• 2.8 mL Blocking Reagent, –20°C
• 120 mL Label Probe Solution, 2–8°C
• 120 mL Pre-Amplifier Solution, 2–8°C
• 120 mL Amplifier Solution, 2–8°C
• 360 μL SAPE, 2–8°C
• 120 mL SAPE Diluent, 2–8°C
• 120 mL Lysis Mixture, 15–30°C
• 8 mL Wash Buffer Component 1, 15–30°C
• 130 mL Wash Buffer Component 2, 15–30°C
• 2 x 700 mL SAPE Wash Buffer, 15–30°C
• 10 Magnetic Separation Plates, 15–30°C
• 45 Plate Seals, 15–30°C
• 10 Hybridization Plates, 15–30°C
• 12 Pressure Seals, 15–30°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text