Invitrogen™

QuantiGene™ Sample Processing Kit, tissues

Catalog number: QS0104
Have Questions?
Invitrogen™

QuantiGene™ Sample Processing Kit, tissues

Catalog number: QS0104
Have Questions?
Catalog Number
QS0104
Unit Size
10 samples
Price (USD)
Quantity
Catalog Number
Unit Size
25 samples
Price (USD)
Quantity
Catalog Number
Unit Size
100 samples
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
QS010410 samples
QS010525 samples
QS0106100 samples
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The QuantiGene Sample Processing Kit for tissues contains reagents and instructions for the preparation of tissue homogenates from fresh or frozen animal or plant tissues for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.

  • Easy to follow instructions
  • Preserves the integrity of the RNA/DNA without degradation
  • Choose the method that matches your tissue sample

What you get

The QuantiGene Sample Processing Kit for tissues contains our proprietary homogenizing solution, proteinase K, and detailed instructions for preparing homogenized tissue from a variety of tissue sources.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Kit
Sample Type
Tissue
Product Line
QuantiGene™
Product Type
Sample Processing Kit
Quantity
10 Samples
Shipping Condition
Blue Ice

Contents & Storage

• 10 mL Homogenizing Solution, store at 15°C to 30°C
• 36 μL Proteinase K (50 μg/μL), store at -20°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text