Invitrogen™

QuantiGene™ Sample Processing Kit, FFPE samples

Catalog number: QS0108
Have Questions?
Invitrogen™

QuantiGene™ Sample Processing Kit, FFPE samples

Catalog number: QS0108
Have Questions?
Catalog Number
QS0108
Unit Size
25 samples
Price (USD)
Quantity
Catalog Number
Unit Size
10 samples
Price (USD)
Quantity
Catalog Number
Unit Size
100 samples
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
QS010825 samples
QS010710 samples
QS0109100 samples
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The QuantiGene Sample Processing Kit for formalin-fixed paraffin-embedded (FFPE) samples contains reagents and instructions for the preparation of tissue homogenates from FFPE tissue samples for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets. H&E stained slides are compatible with QuantiGene assays.

  • Easy to follow instructions
  • Preserves the integrity of the RNA/DNA without degradation
  • Choose the method that matches your FFPE tissue sample

What you get

The QuantiGene Sample Processing Kit for FFPE samples contains our proprietary homogenizing solution, proteinase K, and detailed instructions for preparing tissue homogenates from FFPE tissue sections.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Kit
Sample Type
FFPE
Product Line
QuantiGene™
Product Type
Sample Processing Kit
Quantity
25 Samples
Shipping Condition
Blue Ice

Contents & Storage

• 20 mL Homogenizing Solution, store at 15°C to 30°C
• 90 μL Proteinase K (50 μg/μL), store at -20°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text