Invitrogen™

QuantiGene™ Sample Processing Kit, blood samples

Catalog number: QS0112
Have Questions?
Invitrogen™

QuantiGene™ Sample Processing Kit, blood samples

Catalog number: QS0112
Have Questions?
Catalog Number
QS0112
Unit Size
5 x 10 plates
Price (USD)
Quantity
Catalog Number
Unit Size
2 plates
Price (USD)
Quantity
Catalog Number
Unit Size
10 plates
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
QS01125 x 10 plates
QS01102 plates
QS011110 plates
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

This QuantiGene Sample Processing Kit for blood samples contains reagents and instructions for the preparation of cell lysates from whole blood samples for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.

  • Easy to follow instructions
  • Preserves the integrity of the RNA/DNA without degradation
  • Choose the method that matches your whole blood or dried blood spot sample

What you get

The kit contains our proprietary lysis mixture, proteinase K, and detailed instructions for preparing whole blood from heparin, citrate, or EDTA tubes, PAXgene blood RNA tubes, Tempus™ blood RNA tubes, or from dried blood spots prepared from standard anticoagulant venous blood or finger sticks.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Kit
Sample Type
Blood
Product Line
QuantiGene™
Product Type
Sample Processing Kit
Quantity
5 x 10 Plates
Shipping Condition
Blue Ice

Contents & Storage

• 5 x 50 mL Lysis Mixture, store at 15°C to 30°C
• 5 x 3.25 mL Proteinase K (50 μg/μL), store at -20°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text