Invitrogen™

QuantiGene™ Temperature Validation Kit

Catalog number: QS0517
Have Questions?
Invitrogen™

QuantiGene™ Temperature Validation Kit

Catalog number: QS0517
Have Questions?
Catalog Number
QS0517
Unit Size
1 kit
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
QS05171 kit
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The QuantiGene Incubator Temperature Validation Kit is designed to precisely measure the temperatures in the incubators used in QuantiGene Singleplex and QuantiGene Plex assays. The kit uses a National Institute of Standards and Technology (NIST)-calibrated thermometer to enable users to assess the accuracy of the temperature of ovens or incubators. The kit can be used to monitor incubators used for QuantiGene assays to ensure adherence to recommended incubation conditions.

What you get
The QuantiGene Incubator Temperature Validation Kit contains a QuantiGene mock hybridization 96-well plate, an NIST-calibrated thermometer and thermocouple probe, and a procedure for validating a static or shaking incubator.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
QuantiGene™ Incubator
No. of Places
96
Type
Temperature Validation Kit
Product Line
QuantiGene™
Shipping Condition
Room Temperature

Contents & Storage

• 1 NIST-calibrated digital thermometer
• 1 Sealed hybridization plate

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text