Search by lot number or partial lot number
Catalog Number | Unit Size | Price (USD) | Quantity | |
---|---|---|---|---|
QS0641 | 10 µg | 125.00 |
Universal Rat Reference RNA is an RNA extract used as a positive control in QuantiGene assays. The Universal Rat Reference RNA is composed of total RNA from 14 rat cell lines:
Equal quantities of DNase-treated total RNA from each cell line were pooled to make the universal control.