1x1 image pixel for data collection
ViewRNA™ ISH Cell Assay Kit
ViewRNA™ ISH Cell Assay Kit
Invitrogen™

ViewRNA™ ISH Cell Assay Kit

The ViewRNA™ ISH Cell Assay is a direct fluorescence RNA in situ hybridization method that enables the simultaneous detection ofRead more
Have Questions?
Catalog NumberQuantity
QVC00011 kit
Catalog number QVC0001
Price (USD)
1,925.65
Online Exclusive
2,135.00
Save 209.35 (10%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
1 kit
Price (USD)
1,925.65
Online Exclusive
2,135.00
Save 209.35 (10%)
Each
Add to cart
Ask our AI about this Product
The ViewRNA™ ISH Cell Assay is a direct fluorescence RNA in situ hybridization method that enables the simultaneous detection of one to four RNA targets (either mRNA or non-coding RNA) at single copy sensitivity and single cell resolution using fluorescence microscopes or high-content imagers. Unlike traditional FISH techniques which are generally limited by high background and low sensitivity, the assay uses proprietary chemistry for the target specific probe sets (RNA FISH probes) and branch DNA signal amplification (bDNA) for detection of specific signal. The fluorescence RNA in situ hybridization assay has four main steps: sample preparation, target hybridization, signal amplification, and detection. This ViewRNA ISH assay kit can detect up to three RNA but can be combined with the ViewRNA™ 740 Module to add the fourth probe.

Required additional components:
10X PBS (cat. no. QVC0508)
ViewRNA™ - Detergent Solution QC (cat. No. QVC0509)
ViewRNA™ - Wash Buffer Set (cat. no. QG0507)
ViewRNA™ ISH Cell Accessory Kit (cat. no. QVC0700) intended to provide many of the required components, not supplied in the reagent kit, in order to perform the assay.
ViewRNA™ Probe Sets are not included in the assay and are designed for use with the ViewRNA ISH Cell Assays. Visit our website to view a complete listing of over 6,500 synthesized Probe Sets. By request, new Probe Sets can be designed and synthesized in less than two weeks with no additional costs.

To add the 4th RNA probe:
ViewRNA™ ISH Cell 740 Module (cat. no. QVC0200) designed to be used in conjunction with ViewRNA™ ISH Cell Assay and allows analysis of an additional RNA target in the 740 channel (AlexaFluor 750) See the package insert for a complete list of materials provided in the kit.

Reported Applications
Microscopy

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity1 kit
TypeDetection Assay Kit
Unit SizeEach
Contents & Storage
Components sufficient for 1-plex to 3-plex (AlexaFluor 488, 546 and 647) assay in 24- well plate format (24 samples), 96 assays when using cover slips mounted on glass slides

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

How do ViewRNA assays compare to RNAScope assays?

ViewRNA and RNAScope technologies rely on the same signal amplification strategy - branched DNA amplification. Historically, both ViewRNA and RNAScope technologies originated from the same company, Panomics. The assays are expected to yield similar sensitivity and resolution, however each technology relies on its own set of proprietary reagents and probe set designs. Hence, the assays are not considered interchangeable or compatible. ViewRNA probe sets are not tested for RNAScope assaya and vice versa.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

How is the signal amplified in ViewRNA assays?

The ViewRNA technology relies on branched DNA signal amplification strategy. Target probes complementary to the target transcript sequence are further hybridized with pre-amplifier, amplifier and label probes that consist of branched DNA, and form 'tree branches' that allow numerous label probes to attach. This approach allows higher signal amplification compared to traditional ISH techniques.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Where can I find general information about ViewRNA ISH Assays?

For general information about ViewRNA ISH Assays, please go to this page (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cellular-imaging/in-situ-hybridization-ish/rna-fish/viewrna-assays.html).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

For ViewRNA assays, can I use the same set of wash solutions for all samples, including the positive and negative controls?

We do not recommend doing this. The negative control should be processed and washed separately from the rest of the samples. This is because the negative control does not contain any target probe sets and only the amplification reagents are added to it. If experimental samples are washed in the same beaker of wash solutions as the negative control, any unbound target probes that wash away can carry over to the negative control sample and cause unexpected positive signal (that will also appear to be very specific).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

The ViewRNA ISH Cell Assay Kit user manual lists an accessory kit, Cat. No. QVC0700 which has been discontinued. Do you offer or suggest alternatives for the items in the accessory kit that have also been discontinued?

Please contact our Technical Support team at cellanalysis.support@thermofisher.com for the list of suggested alternatives.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
415954-105Certificate of AnalysisJul 10, 2025QVC0001
415954-305Certificate of AnalysisJul 10, 2025QVC0001
415954-205Certificate of AnalysisJul 10, 2025QVC0001
415954-304Certificate of AnalysisJun 14, 2025QVC0001
415954-104Certificate of AnalysisJun 14, 2025QVC0001
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (27)

Citations & References
Abstract
Raphe serotonin neuron-specific oxytocin receptor knockout reduces aggression without affecting anxiety-like behavior in male mice only.
Authors:Pagani JH, Williams Avram SK, Cui Z, Song J, Mezey É, Senerth JM, Baumann MH, Young WS
Journal:
PubMed ID:25677455
'Serotonin and oxytocin influence aggressive and anxiety-like behaviors, though it is unclear how the two may interact. That the oxytocin receptor is expressed in the serotonergic raphe nuclei suggests a mechanism by which the two neurotransmitters may cooperatively influence behavior. We hypothesized that oxytocin acts on raphe neurons to influence ... More
Frequent proviral integration of the human betaretrovirus in biliary epithelium of patients with autoimmune and idiopathic liver disease.
Authors:Wang W, Indik S, Wasilenko ST, Faschinger A, Carpenter EJ, Tian Z, Zhang Y, Wong GK, Mason AL
Journal:
PubMed ID:25521721
'A human betaretrovirus (HBRV) has been linked with primary biliary cirrhosis (PBC) following the detection of viral particles in biliary epithelium by electron microscopy and cloning of the betaretrovirus genome from biliary epithelium and peri-hepatic lymph nodes. Evidence for viral infection was found in the majority of PBC patients'' peri-hepatic ... More
High-throughput detection of miRNAs and gene-specific mRNA at the single-cell level by flow cytometry.
Authors:Porichis F, Hart MG, Griesbeck M, Everett HL, Hassan M, Baxter AE, Lindqvist M, Miller SM, Soghoian DZ, Kavanagh DG, Reynolds S, Norris B, Mordecai SK, Nguyen Q, Lai C, Kaufmann DE
Journal:
PubMed ID:25472703
Fluorescent in situ hybridization (FISH) is a method that uses fluorescent probes to detect specific nucleic acid sequences at the single-cell level. Here we describe optimized protocols that exploit a highly sensitive FISH method based on branched DNA technology to detect mRNA and miRNA in human leukocytes. This technique can ... More
RNA sequencing of pancreatic circulating tumour cells implicates WNT signalling in metastasis.
Authors:Yu M, Ting DT, Stott SL, Wittner BS, Ozsolak F, Paul S, Ciciliano JC, Smas ME, Winokur D, Gilman AJ, Ulman MJ, Xega K, Contino G, Alagesan B, Brannigan BW, Milos PM, Ryan DP, Sequist LV, Bardeesy N, Ramaswamy S, Toner M, Maheswaran S, Haber DA
Journal:Nature
PubMed ID:22763454
Circulating tumour cells (CTCs) shed into blood from primary cancers include putative precursors that initiate distal metastases. Although these cells are extraordinarily rare, they may identify cellular pathways contributing to the blood-borne dissemination of cancer. Here, we adapted a microfluidic device for efficient capture of CTCs from an endogenous mouse ... More
Urinary Xist is a potential biomarker for membranous nephropathy.
Authors:Huang YS, Hsieh HY, Shih HM, Sytwu HK, Wu CC
Journal:
PubMed ID:25157805
Membranous nephropathy (MN), a type of glomerular nephritis, is the most common cause of nephrotic syndrome in human adults. Changes in gene expression as a result of epigenetic dysregulation through long noncoding RNAs (lncRNAs) are increasingly being recognized as important factors in disease. Using an experimental MN mouse model, we ... More
27 total citations

Share catalog number, name or link