Wash Buffer Components (1 and 2)
Product Image
Invitrogen™

Wash Buffer Components (1 and 2)

Wash Buffer Components (1 and 2) are an individual component of the ViewRNA™ ISH Tissue Assay Kits, 1-Plex (Cat. No.Read more
Have Questions?
Catalog NumberQuantity
QVT050224 Assays
Catalog number QVT0502
Price (USD)
528.00
Each
Add to cart
Quantity:
24 Assays
Price (USD)
528.00
Each
Add to cart

Wash Buffer Components (1 and 2) are an individual component of the ViewRNA™ ISH Tissue Assay Kits, 1-Plex (Cat. No. QVT0050). It is an aqueous buffered solution containing detergent, and can be used for 24 assays (108 mL). Store at -15°C to -30°C.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormulationAqueous buffered solution
Product TypeWash Buffer Component
Quantity24 Assays
Unit SizeEach
Contents & Storage
Wash Buffer Components (1 and 2), 108 mL

Sufficient for 24 assays.
Store at -15°C to -30°C.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link