Search Thermo Fisher Scientific
Catalog Number | Unit Size | Price (USD) | Availability | Quantity | |
---|---|---|---|---|---|
QVT0516 | 1 each | 461.00 | - |
The ViewRNA Tissue Positive Control Kit (2-Plex) is designed for use with the ViewRNA Tissue Assay Core Kit, 1-plex (Cat. No. 19931) or ViewRNA Tissue Assay Blue Module (19932). This positive control kit includes 4-6 μm thick rat kidney FFPE sections attached to positively charged slides as well one vial each of TYPE 1 rat Spp1 and TYPE 6 rat Ubc probe sets.