Invitrogen™

AP Stop QT

Catalog number: QVT0517
Have Questions?
Invitrogen™

AP Stop QT

Catalog number: QVT0517
Have Questions?
Catalog Number
QVT0517
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
QVT051710 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
AP Stop QT, 10 mL, is an aqueous buffered solution intended for the inactivation of residual LP6-AP activity after the Fast Blue Substrate development. It is an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex, 24 assays.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
AP Stop QT Solution
Quantity
10 mL

Contents & Storage

Store at 2°C to 8°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text