Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Color | Green (FITC), Red (Texas Red), Deep Red (Cy5), Near Infrared (Cy7) |
Detection Method | Fluorescence |
Format | Kit |
Label or Dye | Alexa Fluor 488, Alexa Fluor 594, Alexa Fluor 647, Alexa Fluor 750 |
Final Product Type | DNA |
Sample Type | Tissue (FFPE), Tissue (CRYO), Cell Culture Cells |
Labeling Target | RNA (General) |
Excitation/Emission | 496/519, 590/617, 650/665, 749/775 nm |
For Use With (Application) | RNA-FISH |
Labeling Method | ViewRNA Type 1, 4, 6, 10 |
Product Line | ViewRNA™ |
Product Type | Tissue Fluorescence 4 Plex Assay Kit |
Quantity | 96 Slides |
Unit Size | Each 1 |
Color | Green (FITC), Orange (TRITC), Deep Red (Cy5), Near Infrared (Cy7) |
Detection Method | Fluorescence |
Format | Kit |
Label or Dye | Alexa Fluor 488, Alexa Fluor 546, Alexa Fluor 647, Alexa Fluor 750 |
Final Product Type | DNA |
Sample Type | Tissue (FFPE), Tissue (CRYO), Cell Culture Cells |
Labeling Target | RNA (General) |
Excitation/Emission | 496/519, 556/573, 650/665, 749/775 nm |
For Use With (Application) | RNA-FISH |
Labeling Method | ViewRNA Type 1, 4, 6, 10 |
Product Line | ViewRNA™ |
Product Type | Tissue Fluorescence 4 Plex Assay Kit |
Quantity | 24 Slides |
Unit Size | Each 1 |
Color | Green (FITC), Orange (TRITC), Deep Red (Cy5), Near Infrared (Cy7) |
Detection Method | Fluorescence |
Format | Kit |
Label or Dye | Alexa Fluor 488, Alexa Fluor 546, Alexa Fluor 647, Alexa Fluor 750 |
Final Product Type | DNA |
Sample Type | Tissue (FFPE), Tissue (CRYO), Cell Culture Cells |
Labeling Target | RNA (General) |
Excitation/Emission | 496/519, 556/573, 650/665, 749/775 nm |
For Use With (Application) | RNA-FISH |
Labeling Method | ViewRNA Type 1, 4, 6, 10 |
Product Line | ViewRNA™ |
Product Type | Tissue Fluorescence 4 Plex Assay Kit |
Quantity | 96 Slides |
Unit Size | Each 1 |
Color | Green (FITC), Red (Texas Red), Deep Red (Cy5), Near Infrared (Cy7) |
Detection Method | Fluorescence |
Format | Kit |
Label or Dye | Alexa Fluor 488, Alexa Fluor 594, Alexa Fluor 647, Alexa Fluor 750 |
Final Product Type | DNA |
Sample Type | Tissue (FFPE), Tissue (CRYO), Cell Culture Cells |
Labeling Target | RNA (General) |
Excitation/Emission | 496/519, 590/617, 650/665, 749/775 nm |
For Use With (Application) | RNA-FISH |
Labeling Method | ViewRNA Type 1, 4, 6, 10 |
Product Line | ViewRNA™ |
Product Type | Tissue Fluorescence 4 Plex Assay Kit |
Quantity | 24 Slides |
Unit Size | Each 1 |
Color | Deep Red (Cy5) |
Detection Method | Fluorescence |
Format | Kit |
Label or Dye | Alexa Fluor 647 |
Final Product Type | DNA |
Sample Type | Tissue (FFPE), Tissue (CRYO), Cell Culture Cells |
Labeling Target | RNA (General) |
Excitation/Emission | 650/665 nm |
For Use With (Application) | RNA-FISH |
Labeling Method | ViewRNA Type 6 |
Product Line | ViewRNA™ |
Product Type | Tissue Fluorescence 1 Plex Assay Core Kit |
Quantity | 24 Slides |
Unit Size | Each 1 |
Color | Deep Red (Cy5) |
Detection Method | Fluorescence |
Format | Kit |
Label or Dye | Alexa Fluor 647 |
Final Product Type | DNA |
Sample Type | Tissue (FFPE), Tissue (CRYO), Cell Culture Cells |
Labeling Target | RNA (General) |
Excitation/Emission | 650/665 nm |
For Use With (Application) | RNA-FISH |
Labeling Method | ViewRNA Type 6 |
Product Line | ViewRNA™ |
Product Type | Tissue Fluorescence 1 Plex Assay Core Kit |
Quantity | 96 Slides |
Unit Size | Each 1 |
Color | Green (FITC) |
Detection Method | Fluorescence |
Format | Kit |
Label or Dye | Alexa Fluor 488 |
Final Product Type | DNA |
Sample Type | Tissue (FFPE), Tissue (CRYO), Cell Culture Cells |
Labeling Target | RNA (General) |
Excitation/Emission | 496/519 nm |
For Use With (Application) | RNA-FISH |
Labeling Method | ViewRNA Type 4 |
Product Line | ViewRNA™ |
Product Type | Tissue Alexa Fluor 488 (Type 4) Module |
Quantity | 24 Slides |
Unit Size | Each 1 |
Color | Green (FITC) |
Detection Method | Fluorescence |
Format | Kit |
Label or Dye | Alexa Fluor 488 |
Final Product Type | DNA |
Sample Type | Tissue (FFPE), Tissue (CRYO), Cell Culture Cells |
Labeling Target | RNA (General) |
Excitation/Emission | 496/519 nm |
For Use With (Application) | RNA-FISH |
Labeling Method | ViewRNA Type 4 |
Product Line | ViewRNA™ |
Product Type | Tissue Alexa Fluor 488 (Type 4) Module |
Quantity | 96 Slides |
Unit Size | Each 1 |
Color | Orange (TRITC) |
Detection Method | Fluorescence |
Format | Kit |
Label or Dye | Alexa Fluor 546 |
Final Product Type | DNA |
Sample Type | Tissue (FFPE), Tissue (CRYO), Cell Culture Cells |
Labeling Target | RNA (General) |
Excitation/Emission | 556/573 nm |
For Use With (Application) | RNA-FISH |
Labeling Method | ViewRNA Type 1 |
Product Line | ViewRNA™ |
Product Type | Tissue Alexa Fluor 546 (Type 1) Module |
Quantity | 24 Slides |
Unit Size | Each 1 |
Color | Orange (TRITC) |
Detection Method | Fluorescence |
Format | Kit |
Label or Dye | Alexa Fluor 546 |
Final Product Type | DNA |
Sample Type | Tissue (FFPE), Tissue (CRYO), Cell Culture Cells |
Labeling Target | RNA (General) |
Excitation/Emission | 556/573 nm |
For Use With (Application) | RNA-FISH |
Labeling Method | ViewRNA Type 1 |
Product Line | ViewRNA™ |
Product Type | Tissue Alexa Fluor 546 (Type 1) Module |
Quantity | 96 Slides |
Unit Size | Each 1 |
Catalog Number | Specifications | Unit Size | Product Type | Color | Quantity | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
QVT4800 | Each 1 | Tissue Fluorescence 4 Plex Assay Kit | Green (FITC), Red (Texas Red), Deep Red (Cy5), Near Infrared (Cy7) | 96 Slides | 4,968.65 Online price 5,460.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
QVT0700 | Each 1 | Tissue Fluorescence 4 Plex Assay Kit | Green (FITC), Orange (TRITC), Deep Red (Cy5), Near Infrared (Cy7) | 24 Slides | 1,469.65 Online price 1,606.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
QVT4700 | Each 1 | Tissue Fluorescence 4 Plex Assay Kit | Green (FITC), Orange (TRITC), Deep Red (Cy5), Near Infrared (Cy7) | 96 Slides | 4,968.65 Online price 5,460.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
QVT0800 | Each 1 | Tissue Fluorescence 4 Plex Assay Kit | Green (FITC), Red (Texas Red), Deep Red (Cy5), Near Infrared (Cy7) | 24 Slides | 1,461.65 Online price 1,606.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
QVT0600C | Each 1 | Tissue Fluorescence 1 Plex Assay Core Kit | Deep Red (Cy5) | 24 Slides | 1,057.65 Online price 1,162.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
QVT4600C | Each 1 | Tissue Fluorescence 1 Plex Assay Core Kit | Deep Red (Cy5) | 96 Slides | 3,389.65 Online price 3,725.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
QVT0688B | Each 1 | Tissue Alexa Fluor 488 (Type 4) Module | Green (FITC) | 24 Slides | 278.65 Online price 305.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
QVT4688B | Each 1 | Tissue Alexa Fluor 488 (Type 4) Module | Green (FITC) | 96 Slides | 941.65 Online price 1,034.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
QVT0646B | Each 1 | Tissue Alexa Fluor 546 (Type 1) Module | Orange (TRITC) | 24 Slides | 278.65 Online price 305.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
QVT4646B | Each 1 | Tissue Alexa Fluor 546 (Type 1) Module | Orange (TRITC) | 96 Slides | 941.65 Online price 1,034.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
With ViewRNA Tissue assays, you can achieve:
Note: A ViewRNA probe set is required and can be purchased separately
ViewRNA Tissue Assay Fluorescence kits, when combined with a ViewRNA Tissue probe set (purchased separately), enables the detection of a single RNA target in tissues/cells using Alexa Fluor dyes. ViewRNA offers 6500 catalog probe sets. Any more probe sets can be designed at no extra cost.
To meet various experimental needs, the ViewRNA Tissue assay is provided in various kits and modules.
ViewRNA Tissue Fluorescence 4-Plex Assay Kit (green, orange, deep red, and near infrared)
With this kit four RNA targets can be detected using ViewRNA probe sets type 1, 4, 6, and 10 (ViewRNA probe sets are sold separately). This kit utilizes Alexa Fluor 546 for Type 1 probes, Alexa Fluor 488 for Type 4 probes, Alexa Fluor 647 for Type 6 probes, and Alexa Fluor 750 for Type 10 probes. RNA visualization requires FITC/GFP, TRITC/RFP, CY5/Deep Red, and Cy7/NIR microscope filter sets. An additional DAPI filter set is needed for nucleus counterstain with DAPI.
ViewRNA Tissue Fluorescence 4-Plex Assay Kit (green, red, deep red, and near infrared)
With this kit four RNA targets can be detected using ViewRNA probe sets type 1, 4, 6, and 10 (ViewRNA probe sets are sold separately). This kit utilizes Alexa Fluor 594 for Type 1 probes, Alexa Fluor 488 for Type 4 probes, Alexa Fluor 647 for Type 6 probes, and Alexa Fluor 750 for Type 10 probes. RNA visualization requires FITC/GFP, Texas Red, CY5/Deep Red, and Cy7/NIR microscope filter sets. An additional DAPI filter set is needed for nucleus counterstain with DAPI.
ViewRNA Tissue Fluorescence 1-Plex Assay Core Kit, with Alexa Fluor 647 (Type 6) label probe
With this kit, one RNA target can be detected using a Type 6 ViewRNA probe set (ViewRNA probe sets are sold separately). Up to four RNA targets can be detected when combined with ViewRNA Tissue modules (sold separately). RNA visualization requires a Cy5/Deep Red microscope filter set. An additional DAPI filter set is needed for nucleus counterstain with DAPI.
ViewRNA Tissue Alexa Fluor 488 (Type 4) Module
When combined with the ViewRNA Tissue Fluorescence 1-Plex Assay Core Kit (Cat. No. QVT0600) this module can detect an additional RNA target utilizing a Type 4 ViewRNA probe set. This module can be combined with other modules for additional RNA target detection. RNA visualization requires a FITC/GFP filter set.
ViewRNA Tissue Alexa Fluor 546 (Type 1) Module
When combined with the ViewRNA Tissue Fluorescence 1-Plex Assay Core Kit (Cat. No. QVT0600) this module can detect an additional RNA target utilizing a Type 1 ViewRNA probe set. This module can be combined with other modules for additional RNA target detection. RNA visualization requires a TRITC/RFP filter set.
Note: The ViewRNA Tissue Alexa Fluor 546 (Type 1) Module cannot be combined with the ViewRNA Tissue Alexa Fluor 594 (Type 1) Module because both use a Type 1 probe set and will spectrally overlap in most fluorescence microscopes.
ViewRNA Tissue Alexa Fluor 594 (Type 1) Module
When combined with the ViewRNA Tissue Fluorescence 1-Plex Assay Core Kit (Cat. No. QVT0600) this module can detect an additional RNA target utilizing a Type 1 ViewRNA probe set. This module can be combined with other modules for additional RNA target detection. RNA visualization requires a Texas Red filter set.
Note: The ViewRNA Tissue Alexa Fluor 594 (Type 1) Module cannot be combined with the ViewRNA Tissue Alexa Fluor 546 (Type 1) Module because both use a Type 1 probe set and will spectrally overlap in most fluorescence microscopes.
ViewRNA Tissue Alexa Fluor 750 (Type 10) Module
When combined with ViewRNA Tissue Fluorescence 1-Plex Assay Core Kit (Cat. No. QVT0600) this module can detect an additional RNA target utilizing a Type 10 ViewRNA probe set. This module can be combined with other modules for additional RNA target detection. RNA visualization requires a CY7/NIR filter set.