Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Biotin-11-dUTP Solution (1 mM)

Catalog number: R0081
Have Questions?
Thermo Scientific™

Biotin-11-dUTP Solution (1 mM)

Catalog number: R0081
Have Questions?
Catalog Number
R0081
Unit Size
50 µL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
R008150 µL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific Biotin-11-dUTP (biotin-epsilon-aminocaproyl-[5-{3-aminoallyl}-2'-deoxyuridine-5'-triphosphate]) is supplied as 1 mM aqueous solution titrated to pH 7.0 with NaOH and is designed for enzymatic non-radioactive labelling of DNA.

Applications
• Enzymatic non-radioactive labeling of DNA by PCR
• Nick-translation
• cDNA synthesis
• Random primed labeling
• Primer extension

Biotin-11-dUTP can be enzymatically incorporated into DNA with:
Reverse transcriptases
Taq DNA Polymerase
phi29 DNA Polymerase
Klenow Fragment, exo
Klenow Fragment
DNA Polymerase I

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form
Liquid
Polymerase
DNA Polymerase
Labeling Method
Direct Labeling
Label or Dye
Biotin
Quantity
50 μL
Shipping Condition
Dry Ice
For Use With (Application)
cDNA Synthesis, Non-Radioactive Labeling
Concentration
1 mM

Contents & Storage

Biotin-11-dUTP Solution, 1 mM (50 μL)

Store at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text