Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermo Scientific™ Remel Buffered CYE Selective Agar wPolymyxin B, Anisomycin, Vancomycin (BCYE w/PAV) is used for selective isolation of Legionella spp.
Description | Buffered CYE Selective Agar; w/Vancomycin |
Format | 12mm x 85mm Monoplate |
Product Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | dimFormat | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R01341 | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific™ Remel Buffered CYE Selective Agar wPolymyxin B, Anisomycin, Vancomycin (BCYE w/PAV) is used for selective isolation of Legionella spp.