Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermo Scientific™ Remel Buffered CYE Agar (Buffered Charcoal Yeast Extract) (BCYE) is used for the cultivation and isolation of Legionella species.
Description | Buffered CYE Agar (BCYE) |
Format | 12mm x 85mm Monoplate |
Product Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | dimFormat | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R01342 | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Ordering Information:
REF R09300 requires addition of BCYE supplement, REF R45005. Optional antimicrobial supplements are PAC, REF R45006, PAV, REF R45007.