Search Thermo Fisher Scientific
Thermo Scientific™ Remel D/E Neutralizing Broth is used in environmental sampling procedures requiring neutralization of chemical disinfectants to distinguish bacteriostatic and bactericidal activity.