Informational:Informational:Save up to 31% on your order of cell culture products. View promotion details »
Gibco™

Gentamicin/Amphotericin Solution

Catalog number: R01510
Have Questions?
Gibco™

Gentamicin/Amphotericin Solution

Catalog number: R01510
Have Questions?
Catalog Number
R01510
Unit Size
10 x 1 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
R0151010 x 1 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
A sterile, concentrated (500×) antibiotic⁄antimycotic solution of gentamicin and amphotericin B (10 × 1 ml vials⁄pkg). One Gentamicin⁄Amphotericin Solution 10-Pak is enough to supplement ten 500 ml bottles of basal medium.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
Amphotericin B, Gentamicin
Physical Form
Liquid
Recommended Storage

minus 20 degrees C

Sterile Filtered
Yes
Concentration
500 X, 125 μg/mL Amphotericin B, 5 mg/mL Gentamicin
Quantity
10 x 1 mL
Validated Application
Prevention of Cell Culture Contamination
Product Line
Cascade Biologics™
Shipping Condition
Dry Ice

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text