Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Certifications/Compliance | Industrial Reference: BAM, COMPF, SMWW, USP. |
Color | Clear pink |
Description | MacConkey Agar Medium |
Format | 85mm Monoplate |
Manufacturing Origin | United States |
Product Type | Agar |
Fill Volume | 19.5 mL |
Packaging Type | Flow Wrapped - Single Layer |
Quantity | 100/Pk. |
Unit Size | Each |
Certifications/Compliance | Industrial Reference: BAM, COMPF, SMWW, USP. |
Color | Clear pink |
Description | MacConkey Agar Medium |
Format | 85mm Monoplate |
Manufacturing Origin | United States |
Product Type | Agar |
Fill Volume | 19.5 mL |
Packaging Type | Flow Wrapped - Single Layer |
Quantity | 10/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | dimFormat | Quantity | Manufacturing Origin | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R01552 | Each | MacConkey Agar Medium | 100/Pk. | United States | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R01550 | Each | MacConkey Agar Medium | 10/Pk. | United States | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|