Thermo Scientific™

MacConkey Agar with Sorbitol

Catalog number: R01561
Have Questions?
Thermo Scientific™

MacConkey Agar with Sorbitol

Catalog number: R01561
Have Questions?

Selectively detect sorbitol negative Escherichia coli from clinical specimens with Thermo Scientific™ Remel™ MacConkey Agar with Sorbitol prepared media plates. The medium is recommended for the isolation of verotoxin-producing E.coli O157: H7.  Unlike most serotypes of E.coli, verotoxin-producing E.coli O157: H7 does not ferment sorbitol and, therefore, produces colorless colonies. In contrast, most E.coli strains ferment sorbitol and form pink colonies.

 
Catalog Number
R01561
Unit Size
Each
Quantity
100/Pk.
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: BAM, COMPF.
DescriptionMacConkey Agar w/Sorbitol
Format12mm x 85mm Monoplate
Product TypeAgar
Quantity100/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
10/Pk.
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: BAM, COMPF.
DescriptionMacConkey Agar w/Sorbitol
Format12mm x 85mm Monoplate
Product TypeAgar
Quantity10/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
20/Pk.
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: BAM, COMPF.
DescriptionMacConkey Agar w/Sorbitol
Format20mm x 150mm Pour Tube
Product TypeAgar
Quantity20/Pk.
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
R01561Full specifications
Each100/Pk.Request A Quote
Certifications/ComplianceIndustrial Reference: BAM, COMPF.
DescriptionMacConkey Agar w/Sorbitol
Format12mm x 85mm Monoplate
Product TypeAgar
Quantity100/Pk.
Unit SizeEach
R01556Full specifications
Each10/Pk.Request A Quote
Certifications/ComplianceIndustrial Reference: BAM, COMPF.
DescriptionMacConkey Agar w/Sorbitol
Format12mm x 85mm Monoplate
Product TypeAgar
Quantity10/Pk.
Unit SizeEach
R09548Full specifications
Each20/Pk.Request A Quote
Certifications/ComplianceIndustrial Reference: BAM, COMPF.
DescriptionMacConkey Agar w/Sorbitol
Format20mm x 150mm Pour Tube
Product TypeAgar
Quantity20/Pk.
Unit SizeEach
Showing 3 of 3

Escherichia coli O157 is a significant human pathogen and is recognized as a cause of hemorrhagic colitis, an illness characterized by bloody diarrhea, severe abdominal pain, and hemolytic uremic syndrome.

Use Sorbitol MacConkey Agar for selective detection of Escherichia coli O157.

  • Sensitive and specific for E. coli O157 based on sorbitol fermentation.
  • Recommended as a simple, inexpensive, rapid and reliable means of screening for E. coli O157.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

For In Vitro Diagnostic Use.  

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text