Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermo Scientific™ Remel Mueller Hinton Agar with 2% NaCl is for use with antibiotic gradient based systems for determination of methicillin and oxacillin susceptibility of staphylococci.
Description | Mueller Hinton Agar w/2% NaCl |
Format | 12mm x 85mm Monoplate |
Product Type | Agar |
Quantity | 10/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | dimFormat | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R01621 | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific™ Remel Mueller Hinton Agar with 2% NaCl is for use with antibiotic gradient based systems for determination of methicillin and oxacillin susceptibility of staphylococci.